Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
product line
ISOM8™
Quality Segment
assay
(HPLC), ≥98%
form
solid
mol wt
484.69
reaction suitability
reaction type: Arylations, reaction type: Buchwald-Hartwig Cross Coupling Reaction, reaction type: C-X Bond Formation, reaction type: Cross Couplings, reaction type: Fluorinations
mp
170-171 °C (Lit.), 166-170 °C
application(s)
diagnostic assay manufacturing
diagnostic assay manufacturing
life science and biopharma
functional group
phosphine
storage temp.
room temp
SMILES string
COc1ccc(OC)c(c1P(C(C)(C)C)C(C)(C)C)-c2c(cc(cc2C(C)C)C(C)C)C(C)C
InChI
1S/C31H49O2P/c1-19(2)22-17-23(20(3)4)27(24(18-22)21(5)6)28-25(32-13)15-16-26(33-14)29(28)34(30(7,8)9)31(10,11)12/h15-21H,1-14H3
InChI key
REWLCYPYZCHYSS-UHFFFAOYSA-N
General description
Welcome to our ISOM8™ line of high-quality products available and able to scale for manufacturing. Within our ISOM8™ team, we have expertise and documentation to support ISO 9001 industrial and non-GMP pharma production needs, as defined by our M-Clarity™ Program.
Our portfolio of ISOM8™ products and chemistries is ever evolving to support our valued Sigma-Aldrich® customers′ requirements. Please navigate to our documents section to look at dossiers and other quality documentation for this product, provided with support of our Elevate Program.
Application
Notable characteristics & key uses:
- Selectivity: The bulky tert-butyl groups provide exceptional steric control, preventing unwanted side reactions and enhancing selectivity.
- Electronic Properties: Strong electron-donating ability due to alkyl substituents and methoxy groups, stabilizing low-valent metal intermediates.
- Air Stability: Unlike traditional phosphine ligands, tBuBrettPhos demonstrates remarkable resistance to air, moisture, and thermal degradation
- Catalytic Applications: tBuBrettPhos is widely used in various reactions, including cross-coupling reactions, such as Suzuki and Sonogashira reactions, due to its ability tostabilize metal-ligand complexes.
- Cross-Coupling Reactions: tBuBrettPhos enables efficient Suzuki-Miyaura, Buchwald-Hartwig, Heck, and Sonogashira coupling reactions under mild conditions with exceptional selectivity and functional group tolerance, facilitating the formation of carbon-carbon bonds
- Phosphine-Catalyzed Reactions: tBuBrettPhos can act as a catalyst or ligand in various phosphine-catalyzed transformations, enhancing reaction rates and selectivity
- C-H Activation: It has been employed in C-H activation processes, allowing for the functionalization of hydrocarbons under mild conditions.
- Asymmetric Synthesis: tBuBrettPhos is used in asymmetric synthesis to produce chiral molecules, often in conjunction with transition metal catalysts to achieve enantioselectivity.
- Organometallic Chemistry: It plays a role in the stabilization of organometallic complexes, which can be crucial for the development of new catalytic systems.
Recent Studies and Innovations:Selective Monoarylation Catalysis: Pd-methylnaphthyl(MeNAP)-tBuBrettPhos complexes demonstrate exceptional selectivity as pre-catalysts for the monoarylation of ammonia and hydrazine, with theirsterically hindered nature preventing over-arylation and enabling precise control over product formation. (Bingxiang Xue et al., 2024
C-O Cross-Coupling with Fluorinated Alcohols: A highly effective protocol has been developed forcross-coupling (hetero)aryl bromides with fluorinated alcohols using tBuBrettPhos Pd G3 and Cs₂CO₃ in toluene, offering short reaction times, excellent functional group tolerance, and broad compatibility with both electron-rich and electron-poor (hetero)arenes. (Robert Szpera et al., 2020)
Nitroaromatic Synthesis: Under weakly basic conditions, tBuBrettPhos supported palladium catalysis enables the efficient transformation of aryl chlorides, triflates, and nonaflates to nitroaromatics, displaying broad substrate scope and excellent functional group compatibility. (Brett P. et al., 2009)
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description ≥98%, ISOM8™ | description 97% | description ≥98%, ISOM8™ | description ≥98%, ISOM8™ |
| reaction suitability reaction type: Buchwald-Hartwig Cross Coupling Reaction, reaction type: Cross Couplings, reaction type: Arylations, reaction type: C-X Bond Formation, reaction type: Fluorinations | reaction suitability reagent type: ligand | reaction suitability reaction type: Fluorinations, reagent type: ligand, reaction type: Cross Couplings, reaction type: Buchwald-Hartwig Cross Coupling Reaction | reaction suitability reaction type: Hiyama Coupling, reaction type: Sonogashira Coupling, reaction type: Negishi Coupling, reaction type: Buchwald-Hartwig Cross Coupling Reaction, reagent type: ligand, reaction type: Heck Reaction, reaction type: Stille Coupling, reaction type: Suzuki-Miyaura Coupling |
| assay (HPLC), ≥98% | assay 97% | assay ≥98%, (HPLC) | assay ≥98%, (HPLC) |
| functional group phosphine | functional group phosphine | functional group phosphine | functional group phosphine |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
| form solid | form solid | form solid | form solid |
| storage temp. room temp | storage temp. - | storage temp. room temp | storage temp. room temp |
Still not finding the right product?
Explore all of our products under tBuBrettPhos
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


