Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
96 wells | Usually Ships in 4 Weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery) | $737.00 |
About This Item
sample type serum
sample type plasma
$737.00
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type cell culture supernatant(s)
sample type serum
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1.5 pg/mL
standard curve range: 2.05-500 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... KLK14(43847)
General description
Application
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
Please type the word sample in the text box provided for lot number.
1 of 1
This Item | |||
|---|---|---|---|
| description - | description for serum, plasma and cell culture supernatants | description for serum, plasma, cell culture supernatants and urine | description - |
| Gene Information human ... KLK14(43847) | Gene Information human ... KLK12(43849) | Gene Information human ... KLK6(5653) | Gene Information human ... CTSL(1514) |
| species reactivity human | species reactivity human | species reactivity human | species reactivity human |
| input sample type cell culture supernatant(s) | input sample type plasma | input sample type cell culture supernatant(s) | input sample type cell culture supernatant(s) |
| assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% |
| detection method colorimetric | detection method colorimetric | detection method colorimetric | detection method colorimetric |
| technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable |
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




