Skip to Content
MilliporeSigma

Skip To

SML2037

SR2595

≥98% (HPLC)

Synonym(s):

(S)-4′-((5-((1-(4-(tert-butyl)phenyl)ethyl)carbamoyl)-2,3-dimethyl-1H-indol-1-yl)methyl)-[1,1′-biphenyl]-2-carboxylic acid, 4′-[[5-[[[(1S)-1-[4-(1,1-Dimethylethyl)phenyl]ethyl]amino]carbonyl]-2,3-dimethyl-1H-indol-1-yl]methyl]-[1,1′-biphenyl]-2-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
$205.00

About This Item

Empirical Formula (Hill Notation):
C37H38N2O3
CAS Number:
Molecular Weight:
558.71
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

$205.00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to faint brown

solubility

DMSO: 2 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

O=C(C(C=C1)=CC2=C1N(C(C)=C2C)CC(C=C3)=CC=C3C4=C(C(O)=O)C=CC=C4)N[C@H](C(C=C5)=CC=C5C(C)(C)C)C

InChI

1S/C37H38N2O3/c1-23-25(3)39(22-26-11-13-28(14-12-26)31-9-7-8-10-32(31)36(41)42)34-20-17-29(21-33(23)34)35(40)38-24(2)27-15-18-30(19-16-27)37(4,5)6/h7-21,24H,22H2,1-6H3,(H,38,40)(H,41,42)/t24-/m0/s1

InChI key

WKDKAVKAQXVFJV-DEOSSOPVSA-N

Biochem/physiol Actions

Inverse agonist of PPARγ that promoted osteogenic differentiation of isolated bone marrow-derived mesenchymal stem cells
SR2595 is an inverse agonist of PPARG (PPARγ) with an IC50 value of 30 nM. It was found to promote osteogenic differentiation of isolated bone marrow-derived mesenchymal stem cells accompanied by increased expression of bone morphogenetic proteins BMP2 and BMP6. SR2595 showed good pharmacokinetics and can be administered orally. It caused no significant change in insulin sensitivity, fasting insulin levels, food consumption or body weight when administered orally to mice.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1179SML2281SML2874
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

shipped in

wet ice

shipped in

-

shipped in

-

shipped in

-

color

white to faint brown

color

white to beige

color

white to beige

color

white to beige


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters