Skip to Content
MilliporeSigma

Skip To

SML2605

TRPC6 PAM 51164

≥98% (HPLC)

Synonym(s):

N-(2-Chlorophenyl)-2-(4-phenylpiperazin-1-yl)acetamide, N-(2-Chlorophenyl)-4-phenyl-1-piperazineacetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
$64.80
$67.10
25 mg
Check Cart for Availability
$263.00
$223.55

About This Item

Empirical Formula (Hill Notation):
C18H20ClN3O
CAS Number:
Molecular Weight:
329.82
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

$67.10

Web-Only Promotion

Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=CC=C1Cl)CN(CC2)CCN2C3=CC=CC=C3

InChI

1S/C18H20ClN3O/c19-16-8-4-5-9-17(16)20-18(23)14-21-10-12-22(13-11-21)15-6-2-1-3-7-15/h1-9H,10-14H2,(H,20,23)

InChI key

PTAUSEMXADGRBI-UHFFFAOYSA-N

Biochem/physiol Actions

51164 is a neuron penetrant, potent and selective positive allosteric modulator of TRPC6 channels that activates neuronal store-operated calcium entry (nSOCE) in the hippocampal neurons spines. 51164 fully rescues mushroom spines in Aβ42-treated neurons. It restores long-term potentiation (LTP) deficit in 6 months old hippocampal slices from 5xFAD mouse model.
neuron penetrant, potent and selective positive allosteric modulator of TRPC6 channels that activates nSOCE in the hippocampal neurons spines

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T6442SAB4300572AB5574
description

≥98% (HPLC)

description

affinity isolated antibody, lyophilized powder

description

affinity isolated antibody

description

Chemicon®, from rabbit

assay

≥98% (HPLC)

assay

-

assay

-

assay

-

form

powder

form

lyophilized powder

form

buffered aqueous solution

form

-

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-

solubility

DMSO: 2 mg/mL, clear

solubility

-

solubility

-

solubility

-

color

white to beige

color

-

color

-

color

-


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters