Skip to Content
MilliporeSigma

Skip To

SML3170

QPD-Otf

≥98% (HPLC)

Synonym(s):

4-chloro-2-(6-chloro-4-oxo-3,4-dihydroquinazolin-2-yl)phenyl trifluoromethanesulfonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
$123.00
25 mg
Check Cart for Availability
$396.00

About This Item

Empirical Formula (Hill Notation):
C15H7Cl2F3N2O4S
CAS Number:
Molecular Weight:
439.19
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

$123.00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C1NC(C2=C(C=CC(Cl)=C2)OS(=O)(C(F)(F)F)=O)=NC3=CC=C(C=C31)Cl

Biochem/physiol Actions

QPD-OTf is a cell penetrant activity-based fluorogenic substrate for kinesin-1 that apparently binds to the nucleotide-binding site which removes triflate caging causing precipitation and crystallization of fluorescent QPD along microtubule cytoskeleton marking a kinesin-1 path from Golgi apparatus toward the cell periphery. QPD-OTf is not a substrate for Eg5.
cell penetrant activity-based fluorogenic substrate for kinesin-1

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML3542SML3195PZ0375
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

room temp

solubility

DMSO: 2 mg/mL, clear (warmed)

solubility

DMSO: 2 mg/mL, clear (Warmed)

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

color

white to beige

color

white to beige

color

white to beige

color

white to beige


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters