Skip to Content
MilliporeSigma

Skip To

U6510

Ubiquitin-Specific Protease 5 human

recombinant, expressed in Sf9 cells, ≥51% (SDS-PAGE)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 μg
Check Cart for Availability
$650.00

About This Item

NACRES:
NA.54
UNSPSC Code:
12352204
Recombinant:
expressed in Sf9 cells
Concentration:
≥0.20 mg/mL protein (Bradford), ≥51% (SDS-PAGE)

$650.00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in Sf9 cells

Quality Segment

form

buffered aqueous glycerol solution

mol wt

96 kDa

concentration

≥0.20 mg/mL protein (Bradford), ≥51% (SDS-PAGE)

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

General description

Human USP5 (Ubiquitin-Specific Protease 5) or Isopeptidase T (isoT), amino acids 2-858 (end) with an N-terminal FLAG-tag, expressed in a baculovirus-infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Biochem/physiol Actions

The tumor suppressor gene P53 was shown to be a target of ubiquitin-specific protease 5.

Physical form

Solution in 40 mM Tris-HCl, 110 mM NaCl, 2.2 mM KCl, 80 ng/μl Flag peptide, 3 mM DTT, and 20% Glycerol at pH 8.0.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
U6135SRP0279I2411
description

recombinant, expressed in Sf9 cells, ≥51% (SDS-PAGE)

description

recombinant, expressed in Sf9 cells, ≥70% (SDS-PAGE)

description

recombinant, expressed in baculovirus infected insect cells, ≥70% (SDS-PAGE)

description

recombinant, expressed in Sf9 cells, ≥90% (SDS-PAGE)

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

aqueous solution

form

buffered aqueous glycerol solution

concentration

≥0.20 mg/mL protein (Bradford), ≥51% (SDS-PAGE)

concentration

≥0.10 mg/mL protein (Bradford)

concentration

>0.02 mg/mL

concentration

≥0.30 mg/mL protein (Bradford)

recombinant

expressed in Sf9 cells

recombinant

expressed in Sf9 cells

recombinant

expressed in baculovirus infected insect cells

recombinant

expressed in Sf9 cells

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

mol wt

96 kDa

mol wt

128 kDa

mol wt

118 kDa

mol wt

47.6 kDa


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters