MilliporeSigma
Millicell® Cloud Services
Millicell® Cloud Services
Cloud Software for Connected Lab Instrumentation 
Millicell® Cloud Services offer convenient downstream analysis, sorting, and archiving of data obtained from our Connected Lab Instruments including the Scepter™ 3.0 Handheld Cell Counter, the Millicell® DCI Digital Cell Imager, and the Millicell® ERS 3.0 Digital Voltohmmeter.
  • Track and record data using streamlined data management tools.
  • Easily aggregate data from multiple samples, plates, or experiments for comprehensive analysis.
  • Analyze trends with instant access to historical data.
Cloud Services for the Scepter™ 3.0 Handheld Automated Cell Counter
Enhance your data analysis. Millicell® Cloud Services enable you to work anytime, anywhere while providing new and advanced tools and features. Millicell® Cloud Services enable you to work anytime, anywhere while providing new and advanced tools and features for the Scepter™ 3.0 Handheld Cell Counter.
Learn how to update and navigate the latest Scepter™ 3.0 Handheld Cell Counter software to access Millicell® Cloud enhancements.
  • View and download histograms, cell counts, and associated data
  • Recalculate cell concentration with the dilution calculator tool
  • Compare data with histogram overlays
  • Apply multiple gate adjustments for analysis
  • Data management
  • Data export
Cloud Services for the Millicell® DCI Digital Cell Imager
Easily access your data out of the lab for greater flexibility. With Millicell® Cloud Services, images are automatically transferred from the Millicell® DCI instrument to the Cloud via Wi‑Fi®. Data can then be viewed, sorted, and re-analyzed as desired using the web-based application.
Watch this video to learn more about the instrument’s wireless transfer capabilities for off-device visualization, storage, and additional analysis.
  • View and download raw images
  • Mask editing and adjustment
  • Confluency and estimated cell count redetermination
  • Project management and editing
  • Data export
  • Growth trend analysis
Cloud Services for the Millicell® ERS 3.0 Digital Voltohmmeter
Extend your capability. The Millicell® Cloud features comprehensive measurement view, enabling visualization and analysis of data from the Millicell® ERS 3.0 Digital Voltohmmeter in various formats. Filter measurements based on project, cell line, or user. Aggregate data from multiple projects, perform statistical analyses, and view or manage your data from practically anywhere.
Explore advanced features of the Millicell® ERS 3.0 Digital Voltohmmeter enabled by Millicell® Cloud.
  • Graph plotting
  • Heatmap visualization
  • Plate information display
  • Plate editing
  • Plate management
  • Data export
The Millicell® Cloud automatically graphs data from the Millicell® ERS 3.0, averaging measurements within well groupings. Both bar graphs and line graphs can be quickly and easily exported as png files. Pull data from multiple experiments by selecting a device, cell line, project, and plate name to generate an automatic plot.
On left, a bar graph plotting electrical resistance vs. experiment date. Three different seeding densities of each of two cell lines are depicted. On right, the same data are plotted as overlaid line graphs.
Millicell® Cloud TEER Data Graphs. A) Millicell® Cloud bar graph, B) Millicell® Cloud line graph.
Expanded Capability. Easier Access. Enhanced Analysis.
Cloud Services Step 1
Cloud Services Step 2
Cloud Services Step 3
Millicell® Cloud Interface Example Screens
Request Information