MilliporeSigma
HomeTerpenes in Cannabis: Profile and ImportanceHeadspace SPME-GC/MS Analysis of Terpenes in Hops and Cannabis
Headspace SPME-GC/MS Analysis of Terpenes in Hops and Cannabis
Katherine K. Stenerson, Principal Scientist
Article from Analytix Reporter - Issue 3
In this application, headspace-SPME combined with GC/MS was used to analyze some of the terpenes present in both common hops and cannabis.
Terpenes are small molecules synthesized by some plants. The name terpene is derived from turpentine, which contains high concentrations of these compounds. Terpene molecules are constructed from the joining of isoprene units in a head-to-tail configuration (Figure 1). Classification is then done according to the number of these isoprene units in the structure (Table 1). The configurations of terpenes can be cyclic or open, and can include double bonds, and hydroxyl, carbonyl or other functional groups. If the terpene contains elements other than C and H, it is referred to as a terpenoid.1
Isoprene Unit
Figure 1. Isoprene Unit
ClassificationNumber of Isoprene Units
Monoterpene2
Sesquiterpene3
Diterpene4
Triterpene6
Tetraterpene8
Table 1.Classification of Terpenes
Terpenes are present in essential oils derived from plants and often impart characteristic aromas to the plant or its oil. For example, d-Limonene, which is found in lemon, orange, caraway, and other plant oils, has a lemon-like odor. Essential oils, with their component terpenes and terpenoids, have been applied in therapeutic use known as aromatherapy to aid in the relief of conditions such as anxiety, depression, and insomnia.2 This has led to the use of plants which contain these compounds in preparations such as oils, teas, and tonics.
Using Terpene Profile for Plant Identification
The cannabis sativa (cannabis or marijuana) plant contains over 100 different terpenes and terpenoids, including mono, sesqui, di, and tri, as well as other miscellaneous compounds of terpenoid orgin.3 Although the terpene profile does not necessarily indicate geographic origin of a cannabis sample, it can be used in forensic applications to determine the common source of different samples.4 In addition, different cannabis strains have been developed which have distinct aromas and flavors; a result of the differing amounts of specific terpenes present.5 Humulus lupulus (common hops) and cannabis are both members of the family Cannabaceae.6 Consequently, there are similarities in the terpenes each contains. Terpenes give both plant commodities characteristic organoleptic properties and, in the case of cannabis, produce characteristic aromas when the buds are heated or vaporized.7
Experimental
A Dried cannabis sample was obtained courtesy of Dr. Hari H. Singh, Program Director at the Chemistry & Physiological Systems Research Branch of the United States National Institute on Drug Abuse at the National Institute of Health. The extract strain of the sample was not known. Hop flowers of an unknown variety were purchased from an on-line source. Pellets of Cascade and US Golding hop varieties were purchased at a local home-brew supply shop. Chromatographic separation was performed on an Equity®-1 capillary GC column, and identification was done using retention indices and spectral library matches. Final analytical conditions appear in the figures.
Analysis and Quantification of Terpenes by SPME GC-MS
SPME involves the use of a fiber coated with an extracting phase, which is exposed to the sample to extract the target compounds for terpene quantification. The fiber is then directly inserted into the gas chromatograph (GC) injector, where the compounds are desorbed and separated by the GC column. The separated compounds are then detected and identified by the mass spectrometer (MS). The abundance of each terpene in the sample is determined by analyzing the peak areas or heights in the mass chromatogram. The data obtained from the SPME GC-MS analysis is then processed and analyzed to quantify the terpenes present in the sample.
SPME Method Optimization
The SPME method for terpene analysis was developed using a sample of dried hops flowers (0.2 g in a 10 mL vial). The initial SPME parameters were based on previously published work.8 The GC/MS results of this analysis are shown in Figure 2. This initial set of parameters used 100 µm PDMS fiber, a 1 g sample size, and 60-minute equilibration at room temperature prior to extraction. The sample size was then scaled down to 0.2 g, and the equilibration temperature was increased to 40 °C. This increased temperature allowed the equilibration time to be decreased from 60 to 30 minutes without a loss in sensitivity (Figures 3 and 4). The initial extraction time used was 20 minutes, and a shorter extraction time of 10 minutes was evaluated. However, a loss in sensitivity was noted, so the extraction time was maintained at 20 minutes. The DVB/CAR/PDMS fiber was then evaluated (Figure 5). As expected, this fiber extracted more of the lighter compounds, which, by MS spectral match, were identified as short-chain alcohols and acids.
Headspace SPME-GC/MS Analysis of Dried Hops Flowers (100 µm PDMS Fiber, 1 g Sample)
Figure 2. Headspace SPME-GC/MS Analysis of Dried Hops Flowers (100 µm PDMS Fiber, 1 g Sample)
Sample/matrix:1 g ground hop flowers
SPME fiber:100 µm PDMS (57341-U)
Sample equilibration:60 min, room temperature
Extraction:20 min, headspace, 40 °C
Desorption process:3 min, 270 °C
Fiber post bake:3 min, 270 °C
Column:Equity®-1, 60 m x 0.25 mm I.D., 0.25 µm (28047-U)
Oven:60 °C (2 min), 5 °C/min to 275 °C (5 min)
Inj. temp.:270 °C
Detector:MSD
MSD interface:300 °C
Scan range:full scan, m/z 50-500
Carrier gas:helium, 1 mL/min constant flow
Liner:0.75 mm ID SPME
Headspace SPME-GC/MS Analysis of Dried Hops Flowers
Figure 3. Headspace SPME-GC/MS Analysis of Dried Hops Flowers (100 µm PDMS Fiber, 0.2 g Sample)
Conditions same as Figure 2 except:
sample/matrix: 0.2 g ground hop flowers
Headspace SPME-GC/MS Analysis of Dried Hops Flowers, Increased Sample Equilibration Temperature
Figure 4. Headspace SPME-GC/MS Analysis of Dried Hops Flowers, Increased Sample Equilibration Temperature (100 µm PDMS Fiber, 0.2 g Sample)
Conditions same as Figure 2 except:
sample/matrix:0.2 g ground hop flowers
sample equilibration:30 min, 40 °C
Headspace SPME-GC/MS Analysis of Dried Hops Flowers, Increased Sample Equilibration Temperature
Figure 5. Headspace SPME-GC/MS Analysis of Dried Hops Flowers, Increased Sample Equilibration Temperature (DVB/CAR/PDMS Fiber, 0.2 g Sample)
Conditions same as Figure 2 except:
sample/matrix:0.2 g ground hop flowers
SPME fiber:50/30 µm DVB/CAR/PDMS (57298-U)
sample equilibration:30 min, 40 °C
sample/matrix:0.2 g ground hop flowers
sample equilibration:30 min, 40 °C
Identification of Terpenes Using GC/MS
Using the DVB/CAR/PDMS fiber, samples of hops and cannabis were analyzed using the optimized SPME method. Peak identifications were assigned using MS spectral matching against reference spectra in the Wiley and NIST libraries. Confirmatory identification was done based on retention index. Retention indices were calculated for the compounds identified in each sample using an n-alkane standard analyzed under the same GC conditions. This data was compared with published values (Tables 2 and 3), and final identifications were assigned, as shown in Figures 6 and 7.
Terpenes in Hops Samples
For the dried hop flower sample (Figure 5), the terpene profile should have shown a predominance of β-myrcene, humulene, and caryophyllene, which are typical aroma compounds in hops and hop oil.9 While caryophyllene was identified, both β-myrcene and humulene were not present at levels high enough to be detected by a library search. This may be due to the condition of the sample or the actual variety of hops analyzed since terpene profiles are known to vary between different hop varieties.10 The variety of the hop flowers analyzed is unknown, as the identity was not indicated on the packaging. For comparison, samples of two different varieties of pelletized hops were analyzed after grinding.
These samples appeared green in color, and had a much more characteristic hops-like odor than the dried flowers. Analysis of these samples showed a characteristic terpene profile, with high levels of β-myrcene, caryophyllene, and humulene present in both (Figure 6). The SPME method was able to detect differences in the terpene profiles between the two hops varieties. For example, farnesene (peak 18) was identified in the Cascade hops, but was too low to be confirmed in the US Goldings sample. The level of farnesene in Cascade hops is expected to be 3-7% of total oils, while in US Goldings the level should be <1%.13
Terpenes in Cannabis Sample
The terpenes identified in the cannabis sample (Figure 7) are indicated in Table 3. The profile was similar to those found previously in the analysis of dried cannabis.4,8 Peaks 1-27 in Figure 7 (with the exception of peak 7) were monoterpenes and monoterpenoids. The later eluting peaks consisted of sequiterpenes and caryophyllene oxide, which is a sequiterpenoid. The most abundant terpene was caryophyllene. The predominance of this compound could be due to the specific strain of cannabis tested, and/or the nature of the sample tested, which was dried. Previous studies have shown the level of this compound to increase significantly relative to other terepenes and terpenoids with drying.4 Consequently, the levels of the more volatile monoterpenes and terpenoids would be expected to be less, and this was observed to some degree. Among the monoterpenes and terpenoids the most abundant were α-pinene and d-Limonene.
Peak No.RT (min)NameRI (calculated)RI (literature)Reference
18.58Hexanal  —  78011
212.84α-Pinene  939  94211
313.28Camphene  953  95411
413.716-Methyl-5- hepten-2-one  966  96811
514.1β-Pinene  979  98111
614.41β-Myrcene  988  98611
715.32Cymene1018102011
815.65d-Limonene1030103011
915.98β-Ocimene1041103811
1016.72cis-Linalool oxide1066106811
1117.49Linalool1089109211
1221.86Geraniol1239124311
1325.28Geranyl acetate1363136411
1425.85α-Ylangene138413738
1525.97α-Copaene1388139811
1627.22Caryophyllene1437142811
1727.4trans-α- Bergamotene + unknown1445144312
1817.63trans-β-Farnesene145414508
1928.11Humulene1473146511
2028.41γ-Muurolene1484147511
2128.45γ-Selinene1486147212
2228.68Geranyl isobutyrate1495149311
2328.79β-Selinene149914878
2428.94α-Muurolene1505150011
2528.97α-Selinene1507150112
2629.31γ-Cadinene1521151811
2729.37Calamenene1524151811
2829.45Δ-Cadinene1527152411
2930.93Caryophyllene oxide159015848
3031.5Humulene oxide1614159912
Table 2.Terpenes in Hops Pellets Identified by MS Spectral Library Match and Retention Index.
Headspace SPME-GC/MS Analysis of Hops Pellets Using Final Optimized Method
Figure 6. Headspace SPME-GC/MS Analysis of Hops Pellets Using Final Optimized Method
The peak elution order is listed in Table 2.
Conditions same as Figure 2 except:
sample/matrix:0.5 g ground hop flowers (hops pellets)
SPME fiber:50/30 µm DVB/CAR/PDMS (57298-U)
sample equilibration:30 min, 40 °C
Headspace SPME-GC/MS Analysis of Dried Cannabis Using Final Optimized Method
Figure 7. Headspace SPME-GC/MS Analysis of Dried Cannabis Using Final Optimized Method
The peak elution order is listed in Table 3.
Same as  Figure 2  except:
sample/matrix:0.5 g dried, ground cannabis
SPME fiber:50/30 µm DVB/CAR/PDMS (57298-U)
sample equilibration:30 min, 40 °C
Peak No.RT (min)NameRI (calculated)RI (literature)Reference
1  8.57Hexanal  —  —
210.05Hexene-1-ol  —  —
310.892-Heptanone  —  —
412.56α-Thujene  928  93811
512.86α-Pinene + unknown  939  94211
613.27Camphene  953  95411
713.696-Methyl-5- hepten-2-one  966  96811
814.09β-Pinene  979  98111
914.27β-Myrcene  984  98611
1015.09δ-3-Carene1010101512
1115.2α-Terpinene1014101212
1215.29Cymene1018102011
1315.6d-Limonene1028103011
1416.42γ-Terpinene1056105711
1516.6trans-Sabinene hydrate1062107811
1616.72cis-Linalool oxide1066106811
1717.43Linalool1087109211
1818.04d-Fenchyl alcohol1107111011
1918.82trans-Pinocarveol1135113412
2019.59Borneol L1161116411
2119.811,8-Methandien- 4-ol116811738
2219.81p-Cymen-8-ol1168117212
2319.92Terpinene-4-ol1172118511
2420.22α-Terpineol1181118511
2524.2Piperitenone1322132012
2624.76Piperitenone oxide1344135212
2725.85α-Ylangene138413738
2825.97α-Copaene1388139811
2926.76γ-Caryophyllene1419140312
3027.01α-Santalene1429142812
3127.16Caryophyllene1435142811
3227.36trans-α- Bergamotene + unknown1443144312
3327.49α-Guaiene144814418
3427.56trans-β-Farnesene1451144612
3527.98Humulene1467146511
3628.17Alloaromadendrene1475147811
3728.25α-Curcumene1478147912
3828.75β-Selinene149714878
3928.97α-Selinene150714978
4028.97β-Bisobolene150715068
4129.13α-Bulnesene1514151312
4230.12Selina-3,7(11)- diene1556154212
4330.94Caryophyllene oxide1590159512
4431.5Humulene oxide1614159912
4532.48Caryophylla-3, 8(13)-dien-5-ol A1658165612
Table 3.Terpenes in Dried Cannabis Identified by MS Spectral Library Match and Retention Index
Conclusion
A simple headspace SPME-GC/MS method was used in the analysis of the terpene/terpenoid profiles of both hops and cannabis. The method was able to detect the characteristic terpenes and terpenoids of both, and to distinguish between different hops varieties.
Related Products
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
References
1.
Fessenden R, Fessenden J. 1979. Lipids and Related Natural Products. Organic Chemistry. 2nd Edition. Boston, MA: Willard Grant Press.
2.
Turner CE, Elsohly MA, Boeren EG. 1980. Constituents of Cannabis sativa L. XVII. A Review of the Natural Constituents. J. Nat. Prod.. 43(2):169-234. https://doi.org/10.1021/np50008a001
3.
Hood L, Barry G. 1978. Headspace volatiles of marihuana and hashish: Gas chromatographic analysis of samples of different geographic origin. Journal of Chromatography A. 166(2):499-506. https://doi.org/10.1016/s0021-9673(00)95633-4
4.
Rahn B. 2014. Terpenes: The Flavors of Cannabis Aromatherapy.. [Internet].[cited 15 Sep 2015]. Available from: https://www.leafly.com/news
5.
Plants Database. [Internet].[cited 11 Sep 2015]. Available from: https://plants.sc.egov.usda.gov/java/
6.
2015. Cannabis Safety. Chem. Eng. News Archive. 93(16):27-28. https://doi.org/10.1021/cen-09316-scitech1
7.
Marchini M, Charvoz C, Dujourdy L, Baldovini N, Filippi J. 2014. Multidimensional analysis of cannabis volatile constituents: Identification of 5,5-dimethyl-1-vinylbicyclo[2.1.1]hexane as a volatile marker of hashish, the resin of Cannabis sativa L.. Journal of Chromatography A. 1370200-215. https://doi.org/10.1016/j.chroma.2014.10.045
8.
Wolfe PH, Thesis M. 2012. Oregon State University. [Internet].
9.
Hop varieties. [Internet].[updated 12 Sep 2015]. Available from: https://www.barthhaas.com/
10.
Jennings W, Shibamoto T. 1980. Qualitative Analysis of Flavor and Fragrance Volatiles by Glass Capillary Gas Chromatography. Academic Press.
11.
NIST Chemistry WebBook. [Internet]. Available from: https://webbook.nist.gov/chemistry/
12.
Variety Manual USA Hops. Hop Growers of America. [Internet].[cited 19 Sep 2015]. Available from: https://www.usahops.org/