Graphene Inks for Printed Electronics
By: Ethan B. Secor and Professor Mark C. Hersam,
Department of Materials Science and Engineering and Department of Chemistry, Northwestern University, Evanston, IL 60208-3108 USA
The emerging field of printed electronics requires a suite of functional materials for applications including flexible and large-area displays,1 radio frequency identification tags,2 portable energy harvesting and storage,3,4  biomedical and environmental sensor arrays,5,6 and logic circuits.7 To enable these technologies, functional materials must be integrated with suitable patterning technologies, such as inkjet, gravure, and flexographic printing.8,9 Because electrical conductors are a core component of electronic devices, significant effort has been devoted to conductive materials in the field of printable inks.10 Common conductive inks can be classified into three categories: noble metals, conductive polymers, and carbon nanomaterials.11 This overview focuses on a subset of carbon nanomaterial conductive inks based on solution-processed graphene.
Conductive Inks
The different classes of conductive inks offer unique properties suitable for particular applications. Among the noble metals, silver is the most prevalent printed conductor due to its high conductivity and oxidation resistance, and can be printed using inks based on either silver nanoparticles (Aldrich Prod. Nos. 736465, 736473, 736481, 736503, and 736511) or silver precursors (Aldrich Prod. No. 745707).12,13 These inks offer the highest conductivity among printed materials, but are based on expensive precursors. Copper inks have also been introduced, but typically require core-shell nanoparticle designs or specialized photonic annealing treatments to produce conductive patterns.11 Conductive polymers, such as PEDOT:PSS (Aldrich Prod. Nos. 739316, 768650, 768642), have also been developed for printed electronics applications. These materials offer a modest conductivity at low cost, but are limited in terms of chemical and thermal stability. Carbon nanomaterials, including carbon nanotubes (Aldrich Prod. Nos., 791490, 791504 and 792462) and graphene, offer a low-cost alternative with excellent environmental stability and desirable conductivity, along with unique properties suitable for a range of applications.11
Carbon nanomaterials offer a number of opportunities for printed and flexible electronics. The electrical properties resulting from the sp2-bonded structure of fullerenes, carbon nanotubes, and graphene are particularly promising, and have been exploited in a number of applications from thin-film transistors (TFTs) and electrochemical sensors to supercapacitors and photovoltaics.14,15 Graphene is the two-dimensional sp2-bonded allotrope of carbon equivalent to a single layer of graphite, shown in Figure 1a. With high charge carrier mobility, superlative thermal and chemical stability and intrinsic flexibility, graphene has been demonstrated for a number of applications in printed electronics including chemical and thermal sensors,16,17 micro-supercapacitors,18 and thin-film transistors.19,20 A fundamental challenge to integrating carbon nanomaterials with conventional printing technologies is the production of inks suitable for various deposition processes. Here, we present recent progress in the development of graphene inks using a polymer stabilizer to enable stable, high-concentration inks of pristine graphene with tunable viscosity and solvent composition.
Graphene Production
A number of strategies have been demonstrated to produce graphene including bottom-up techniques such as chemical vapor deposition and epitaxial growth as well as top-down methods such as micromechanical exfoliation and solution-phase exfoliation.21  Among these approaches, the last method offers the most cost-effective and scalable option for large-volume graphene production. The most prevalent methods for graphite exfoliation employ oxidizing agents to produce graphene oxide; however, these strategies lead to degradation of electronic properties.22 On the other hand, pristine graphene can be exfoliated directly by ultrasonication or shear mixing of graphite in organic solvents,23,24 but the most suitable solvents, such as N-methyl pyrrolidone (NMP) and dimethyl formamide (DMF), are expensive, chemically harsh, and difficult to remove.
An alternative technique highlighted here uses the polymer ethyl cellulose (EC) in the inexpensive and benign solvents, such as ethanol.25 This method allows for scalable and efficient production of graphene that is well-suited for solution-phase patterning techniques. The polymer stabilizer enables stable, high-concentration graphene dispersions in a range of solvents including ethanol, terpineol, and cyclohexanone. Consequently, this chemistry offers a general approach to ink development with broadly tunable ink characteristics such as solids loading, viscosity, surface tension, and evaporation kinetics to meet the specific requirements of various printing technologies.26 The graphene flakes produced by this method are characterized by a typical thickness of 1-3 nm and lateral dimension of 50-100 nm.27 A representative atomic force microscopy (AFM) image of the graphene flakes and corresponding particle size distributions are shown in Figure 1b-d. Building on this material, inks can be developed for various printing processes, as will be discussed below.
Solution processed graphene for printed electronics
Figure 1. Solution processed graphene for printed electronics. (a) Schematic graphene structure, highlighting several properties relevant to printed electronics. (b) AFM image of solution-processed graphene flakes. (c) Distribution of flake thicknesses, showing a typical flake thickness of 1-3 nm. (d) Distribution of flake areas, showing a typical flake area of 400-40000 nm2.
Inkjet Printable Graphene Ink (793663)
Piezoelectric inkjet printing offers a promising method for rapid prototyping and fabrication of printed electronic devices due to its non-contact, digital nature and broad materials compatibility. Inkjet printing of pristine graphene is an important capability for integrating graphene into printed devices, and has been reported using two general approaches: dispersing graphene directly in NMP with no binder20,28 and using EC to stabilize graphene in a variety of solvents.18,27 Key results using both of these methods are listed in Table 1. While binder-free dispersion of graphene reduces the requirement for thermal annealing, it offers less versatility to tailor the ink properties such as viscosity and surface tension. Using EC, the solvent system can be tuned to produce stable, high-concentration graphene inks that produce patterns with excellent morphology. Representative images of such patterns are shown in Figure 2, including lines following a single printing pass and dense graphene lines generated from multiple-pass printing. These lines exhibit no evidence of coffee ring formation, and have a reasonable resolution of approximately 60 µm. These patterns were generated using a Fujifilm Dimatix Materials Printer (DMP-2800) equipped with a 10 pL cartridge (DMC-11610) on a SiO2/Si wafer treated with hexamethyldisilazane (HMDS). Importantly, similar patterns can be attained on untreated and O2 plasma-treated SiO2/Si wafers, glass slides, and Kapton™ films.
Table 1. Inkjet printing of pristine graphene
Chemistry Conductivity (S/m) Max Temp. (°C) Graphene Conc. (mg/mL) Resolution (µm) Ref. No.
No binder NMP 100 170 0.11 90 20
Binder: EC Cyclohexanone/Terpineol 25000 250 3.4 60 27
Binder: EC Ethanol/Terpineol 340028 375 1.2 60 18
No binder NMP 3000 70 1.55 80 28
Morphology of inkjet printed graphene features on HMDS-treated Si/SiO2
Figure 2. Morphology of inkjet printed graphene features on HMDS-treated Si/SiO2. Scanning electron micrographs of (a) multiple printed lines and (b) a single printed line and drop (inset, scale bar corresponds to 40 μm) illustrate the uniformity of the printed features. (c) SEM image of a graphene line printed with multiple passes and close-up image showing the resulting dense film microstructure (inset, scale bar = 1 µm). (d) Film cross-section as measured by AFM, averaged over approximately 20 µm of line length for 1, 3, and 10 printing passes. Parts (a,b,d) are adapted with permission from Ref. 27. Copyright 2013, American Chemical Society.
Gravure Printable Graphene Ink (796115)
Gravure printing is ideally suited to high-throughput, roll-to-roll patterning for large-scale production of printed electronics.29 Inks for gravure printing require a high solids loading and well-controlled viscosity and surface tension. Graphene inks have been developed for gravure printing based on EC and terpineol.30 With a solids loading of 10% wt. and a viscosity of 0.75-3 Pa·s, this ink has been printed using a direct gravure printing system to produce patterns with a resolution of approximately 30 µm, as shown in Figure 3. The graphene patterns produced by this method exhibit a film morphology and conductivity that is similar to inkjet-printed patterns. The rheological properties of graphene inks developed for inkjet and gravure printing are listed in Table 2.
Characterization of gravure printed graphene lines
Figure 3. Characterization of gravure printed graphene lines. (a) Line width and (b) line thickness for varying cell size, measured prior to annealing by optical microscopy and optical profilometry, respectively. (c) Large-area scanning electron micrograph of printed lines. (d) Line height profiles measured prior to annealing by optical profilometry, averaged over approximately 2 mm of line length. Reproduced with permission from Ref 30. Copyright 2014, Wiley-VCH Verlag GmbH & Co. KGaA.
Table 2. Graphene inks for inkjet and gravure printing
Product No. Chemistry Printing Method Solids Loading Viscosity (Pa·s) Surface Tension (dyne/cm) Ref. No.
793663 Binder: EC Cyclohexanone/Terpineol Inkjet 2.4% wt. 0.008-0.015 30-35 27
796115 Binder: EC Terpineol Gravure 10% wt. 0.75-3 30-35 30
Graphene Film Characteristics
The properties of solution-processed graphene films depend on the microstructure of the graphene flake network. The two-dimensional structure of graphene is well-suited for producing dense films, but flake aggregation during solvent evaporation can prevent the formation of a uniform film. The inkjet and gravure inks highlighted previously mitigate this problem through the use of the EC binder. This binder effectively isolates the flakes as the ink dries, so the as-deposited films must be thermally annealed to exhibit optimal conductivity. As shown in Figure 4, an annealing treatment in air at 250 °C results in patterns with a high conductivity of approximately 25000 S/m, even for patterns as thin as 150 nm.
Electrical characteristics of graphene films
Figure 4. Electrical characteristics of graphene films. (a) Resistivity of graphene films with different annealing temperatures (30 minutes annealing). (b) Resistivity of graphene films annealed at 250 °C for various times. (c) Resistivity of inkjet-printed graphene lines plotted against line thickness. Adapted with permission from Ref. 27. Copyright 2013, American Chemical Society.
In addition to high electrical conductivity, the printed graphene films exhibit a robust tolerance to bending stresses. As shown in Figure 5, graphene lines printed on Kapton™ showed no measureable change in resistance while strained (tensile strain) to a radius of 4 mm.27 In addition, the lines also retain their electrical properties when subjected to 1000 bending cycles at a radius of curvature of <1 mm. Finally, graphene lines exhibited only a small increase in resistance when folded.
Flexibility characteristics of printed graphene lines on Kapton™ substrates
Figure 5. Flexibility characteristics of printed graphene lines on Kapton™ substrates. (a) Resistance of graphene lines folded to a radius of curvature of 0.9 and 3.4 mm, normalized to the resistance prior to bending. (b) Normalized resistance of graphene lines measured in a flexed state for various degrees of bending, showing reliable retention of electrical conductivity across all measured flex states. (c) Normalized resistance of graphene lines while measured in a folded state, showing a small and irreversible increase in resistance following folding. Images of the sample in the (d) original and (e) folded state. Reproduced with permission from Ref. 27. Copyright 2013, American Chemical Society.
Applications of Printed Graphene
Graphene has been employed for a variety of applications in printed and flexible electronics due to its unique structure and properties. With a conductivity of approximately 25000 S/m, the graphene films described here offer electrical properties comparable to conductive polymer blends while offering a unique set of stability and materials compatibility characteristics. This combination of properties is particularly relevant for thin-film applications, such as contact modification for organic TFTs (OTFTs) and corrosion-resistant conductive coatings. Several studies have demonstrated enhanced charge injection for OTFTs using reduced graphene oxide (RGO) electrodes due to the induced morphology of the semiconductor on the electrode surface.19,31 In addition, graphene electrodes have been employed in carbon nanotube TFTs with promising results.32 Graphene also offers unique characteristics, such as a high surface area, for electrochemical applications including chemical and biological sensors and supercapacitors.21 A brief overview of demonstrated applications for solution-processed graphene is included in Table 3, while a more comprehensive review can be found elsewhere.21
Table 3. Applications of printed graphene
Application Method Material Ref. No.
Thin film transistor Inkjet Printing Graphene 20
Transparent electrode Inkjet Printing Graphene/EC 18
Supercapacitor Inkjet Printing Graphene/EC 18
Gas sensor Inkjet Printing RGO 16, 33
Strain sensor Gravure Printing RGO 34
Temperature sensor Inkjet Printing RGO 17
Acoustic actuator Inkjet Printing RGO 35
Radio frequency antenna Inkjet Printing RGO 36
Resistor Inkjet Printing Graphene/EC 18
OTFT electrode Inkjet Printing RGO/PVA 19
We have shown that stable, high concentration graphene inks can be developed using a stabilizing polymer system. This material system enables broad tuning of the rheology and other fluid properties to realize inks for inkjet and gravure printing. In addition, flake aggregation is mitigated to yield dense and highly conductive graphene films following thermal annealing. The ability to pattern graphene films using solution-based printing techniques enables a number of applications from conductive and stable surface coatings to electrodes for electrochemical sensors, thin-film transistors, and other printed devices.
This work was supported by the Office of Naval Research MURI Program (N00014-11-1-0690).  EBS was further supported by the Department of Defense (DoD) through the National Defense Science and Engineering Graduate (NDSEG) Fellowship Program.
  1. Wood, V.; Panzer, M. J.; Chen, J.; Bradley, M. S.; Halpert, J. E.; Bawendi, M. G.; Bulović, V. Adv. Mater. 2009, 21, 2151.
  2. Subramanian, V.; Fréchet, J. M. J.; Chang, P. C.; Member, S.; Huang, D. C.; Lee, J. B.; Molesa, S. E.; Murphy, A. R.; Redinger, D. R.; Volkman, S. K. Proc. IEEE 2005, 93, 1330.
  3.  Gaikwad, A. M.; Whiting, G. L.; Steingart, D. A.; Arias, A. C. Adv. Mater. 2011, 23, 3251.
  4. Hoth, C. N.; Choulis, S. A.; Schilinsky, P.; Brabec, C. J. Adv. Mater. 2007, 19, 3973.
  5. Jang, J.; Ha, J.; Cho, J. Adv. Mater. 2007, 19, 1772.
  6. Sokolov, A. N.; Roberts, M. E.; Bao, Z. Mater. Today 2009, 12, 12.
  7. Ng, T. N.; Schwartz, D. E.; Lavery, L. L.; Whiting, G. L.; Russo, B.; Krusor, B.; Veres, J.; Bröms, P.; Herlogsson, L.; Alam, N.; Hagel, O.; Nilsson, J.; Karlsson, C. Sci. Rep. 2012, 2, 585.
  8. Arias, A. C.; MacKenzie, J. D.; McCulloch, I.; Rivnay, J.; Salleo, A. Chem. Rev. 2010, 110, 3.
  9. Lupo, D.; Clemens, W.; Breitung, S.; Hecker, K. In Applications of Organic and Printed Electronics; Cantatore, E., Ed.; Springer US: Boston, MA, 2013; pp. 1–26.
  10. Cummins, G.; Desmulliez, M. P. Y. Circuit World 2012, 38, 193.
  11. Kamyshny, A.; Magdassi, S. Small 2014, DOI: 10.1002/smll.201303000.
  12.  Fuller, S. B.; Wilhelm, E. J.; Jacobson, J. M. J. Microelectromechanical Syst. 2002, 11, 54.
  13. Walker, S. B.; Lewis, J. A. J. Am. Chem. Soc. 2012, 134, 1419.
  14. Rouhi, N.; Jain, D.; Burke, P. J. ACS Nano 2011, 5, 8471.
  15. Weiss, N. O.; Zhou, H.; Liao, L.; Liu, Y.; Jiang, S.; Huang, Y.; Duan, X. Adv. Mater. 2012, 24, 5782.
  16. Dua, V.; Surwade, S. P.; Ammu, S.; Agnihotra, S. R.; Jain, S.; Roberts, K. E.; Park, S.; Ruoff, R. S.; Manohar, S. K. Angew. Chem. Int. Ed. Engl. 2010, 49, 2154.
  17. Kong, D.; Le, L. T.; Li, Y.; Zunino, J. L.; Lee, W. Langmuir 2012, 28, 13467.
  18. Li, J.; Ye, F.; Vaziri, S.; Muhammed, M.; Lemme, M. C.; Östling, M. Adv. Mater. 2013, 25, 3985.
  19. Lim, S.; Kang, B.; Kwak, D.; Lee, W. H.; Lim, J. A.; Cho, K. J. Phys. Chem. C 2012, 116, 7520.
  20. Torrisi, F.; Hasan, T.; Wu, W.; Sun, Z.; Lombardo, A.; Kulmala, T. S.; Hsieh, G.-W.; Jung, S.; Bonaccorso, F.; Paul, P. J.; Chu, D.; Ferrari, A. C. ACS Nano 2012, 6, 2992.
  21. Guo, S.; Dong, S. Chem. Soc. Rev. 2011, 40, 2644.
  22. Mao, S.; Pu, H.; Chen, J. RSC Adv. 2012, 2, 2643.
  23. Hernandez, Y.; Nicolosi, V.; Lotya, M.; Blighe, F. M.; Sun, Z.; De, S.; McGovern, I. T.; Holland, B.; Byrne, M.; Gun’Ko, Y. K.; Boland, J. J.; Niraj, P.; Duesberg, G.; Krishnamurthy, S.; Goodhue, R.; Hutchison, J.; Scardaci, V.; Ferrari, A. C.; Coleman, J. N. Nat. Nanotechnol. 2008, 3, 563.
  24. Paton, K. R.; Varrla, E.; Backes, C.; Smith, R. J.; Khan, U.; O’Neill, A.; Boland, C.; Lotya, M.; Istrate, O. M.; King, P.; Higgins, T.; Barwich, S.; May, P.; Puczkarski, P.; Ahmed, I.; Moebius, M.; Coleman, J. N. Nat. Mater. 2014, DOI: 10.1038/NMAT3944.
  25. Liang, Y. T.; Hersam, M. C. J. Am. Chem. Soc. 2010, 132, 17661.
  26. Derby, B. Annu. Rev. Mater. Res. 2010, 40, 395.
  27. Secor, E. B.; Prabhumirashi, P. L.; Puntambekar, K.; Geier, M. L.; Hersam, M. C. J. Phys. Chem. Lett. 2013, 4, 1347.
  28. Finn, D.; Lotya, M.; Cunningham, G.; Smith, R.; McCloskey, D.; Donegan, J.; Coleman, J. N. J. Mater. Chem. C 2013, 925.
  29. Søndergaard, R. R.; Hösel, M.; Krebs, F. C. J. Polym. Sci. Part B Polym. Phys. 2013, 51, 16.
  30. Secor, E. B.; Lim, S.; Zhang, H.; Frisbie, C. D.; Francis, L. F.; Hersam, M. C. Adv. Mater. 2014, DOI: 10.1002/adma.201401052.
  31. Becerril, A.; Stoltenberg, R. M.; Tang, M. L.; Roberts, M. E.; Liu, Z.; Chen, Y.; Kim, D. H.; Lee, B.; Lee, S.; Bao, Z. ACS Nano 2010, 4, 6343.
  32. Su, Y.; Du, J.; Sun, D.; Liu, C.; Cheng, H. Nano Res. 2013, 6, 842.
  33. Huang, L.; Huang, Y.; Liang, J.; Wan, X.; Chen, Y. Nano Res. 2011, 4, 675.
  34. Huang, L.; Wang, Z.-P.; Pu, J.-L.; Shen, L.; Zhang, J.-K. 2013 Seventh Int. Conf. Sens. Technol. 2013, 387.
  35. Shin, K.-Y.; Hong, J.-Y.; Jang, J. Chem. Commun. (Camb). 2011, 47, 8527.
  36. Shin, K.-Y.; Hong, J.-Y.; Jang, J. Adv. Mater. 2011, 23, 2113.
Related Links