MilliporeSigma
HomeUHPLC Analysis Cannabinoids Ascentis Express
UHPLC Analysis Cannabinoids Ascentis Express
UHPLC Analysis of Cannabinoids on Ascentis Express C18
Figure 1.
Conditions
InstrumentShimadzu Nexera X2
ColumnAscentis Express C18, 15 cm x 3.0 mm I.D., 2.7 µm 
Mobile Phase[A] water (0.1% formic acid); [B] acetonitrile (0.085% formic acid) 
Gradient70% B to 88% B in 6 min 
Flow Rate1.0 mL/min 
Pressure5075 psi 
Column Temp.30 °C
DetectorUV, 220 nm
Injection0.6 µL
SampleMixture of 14 cannabinoids, 0.04 mg/mL, 50:50 water:acetonitrile
Table 1
Description
Analysis NoteAs the popularity of cannabis-infused products increases, there is a growing need to characterize the type and content of the cannabinoids found in the product. This application demonstrates the ability of the Ascentis Express C18 column to baseline resolve 14 structurally-similar cannabinoids, in under seven minutes, with excellent peak shape.
Legal InformationAscentis is a trademark of Sigma-Aldrich Co. LLC
Snap-N-Spike is a trademark of Cerilliant Corporation
Snap-N-Shoot Standards is a trademark of Cerilliant Corporation
Application NoMUR110618A
Table 2
Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500