MilliporeSigma
HomeSmall Molecule HPLCHow to Select the Right Water Purification System for Your Lab
How to Select the Right Water Purification System for Your Lab
Stéphane Mabic, PhD
Lab Water Solutions, MilliporeSigma, Guyancourt, France
A laboratory scene showing two scientists in white lab coats working with lab equipment, including two Q-POD ultrapure water dispenser and various glassware.
Purified water is a ubiquitous reagent in most of the analyses performed in laboratories. Because you need to focus on obtaining accurate and reproducible results, it’s important to ensure your lab is equipped with a water purification system that delivers consistently high-quality water meeting all your purity needs. Special care must be taken to select the correct water quality since contaminants such as heavy metals, organics, chlorine or bacteria may impact analyses.
Considerations when choosing a lab water system
Choosing a water purification system for a laboratory can sometimes seem like a complicated process. However, by taking the time to thoroughly evaluate your situation at the beginning of your search and considering a few key parameters, you can make the selection a little easier and have greater confidence in your final choice.
Below are parameters every lab should consider when selecting a solution to deliver purified water in the laboratory:
  • Required water quality
  • Total volume needs
  • Monitoring and traceability
  • Service requirements
  • Certification and compliance
  • Sustainability
  • Design of the building and lab
Investigate your overall lab needs
Identify the different situations in which purified water is used in your laboratory and for which specific applications. Ask yourself these questions:
  • Do you need several different water qualities to accommodate different end purposes (e.g. equipment feed vs. sensitive analyses)?
  • Do you need water feed or points of dispensers at different locations in the lab?
  • What is the average total volume of water you require in a day?
  • How much water is required during hours of peak demand?
Space considerations are also important, as some systems have a variety of possible configurations (under the bench, on the wall, etc.) and can have several point of delivery locations. Such installation options can help you to optimize lab space by uncluttering benchtops and creating a more efficient working environment.
Determine your water quality needs
It's important to know that several types of laboratory water exist. These water grades are defined by standards, which help ensure not only that the correct water type is used for a specific application, but also that cost considerations are taken into account, as Type 1 ultrapure water is more expensive to produce than Type 2 or Type 3 pure water.
With this in mind, identify the different situations in which purified water is used in your laboratory and for which applications.
  • Type 1 water is required for critical laboratory applications, including mobile phase preparation for HPLC or IC; preparation of blanks; standard and sample dilution in HPLC, IC, GC, AA, ICP-MS, etc.; buffer and cell culture preparation for mammalian cell culture; reagent production in molecular biology applications; and preparation of solutions (electrophoresis and blotting).
  • Type 2 water is used in more general applications such as buffer and microbiological culture media preparation, as feed to other laboratory systems (Type 1 water systems, clinical analyzers, dish washers, cell culture incubators and climatic test equipment).
  • Type 3 water is the lowest laboratory water grade and is used in glassware rinsing, heating baths and autoclaves, or also as feed to Type 1 lab water systems.
Depending on your situation, you will need to select a system that can remove water contaminants and produce pure or ultrapure water, or both. As no one technology can remove all contaminants, several purification technologies are generally combined to produce pure and ultrapure water (Figure 1 and Table 1). During your evaluation process, pay careful attention to the technologies used by the systems to make sure you will be able to consistently achieve the water quality needed over time, while also minimizing operating costs.
 Flow schematic of a pure and ultrapure water purification system.
Figure 1.Flow schematic of a pure and ultrapure water purification system.
 IonsOrganicsBacteriaParticulates
Activated Carbon ⬤ ⬤ ⬤ ⬤  
Ion-exchange resins⬤ ⬤ ⬤ ⬤⬤ ⬤  
Reverse osmosis⬤ ⬤ ⬤⬤ ⬤ ⬤⬤ ⬤ ⬤⬤ ⬤ ⬤
Electrodeionization⬤ ⬤ ⬤ ⬤⬤ ⬤  
Bactericidal UV  ⬤ ⬤ ⬤ ⬤ 
Photo-oxidative UV ⬤ ⬤ ⬤  
Microporous filtration  ⬤ ⬤ ⬤ ⬤⬤ ⬤ ⬤ ⬤
Table 1.Common purification technologies and their effectiveness in removing major contaminants.
Evaluate your total water volume requirements
Based on the list of your purified water points of use and applications, now calculate approximately how much water you require per day for each water type. This will provide you with the basic information you will need to discuss recommended installation possibilities with your water purification system vendor. In your estimate, don't forget to take into account days when greater volumes of water may be required.
Understand the importance of monitoring and traceability
It is essential to know that your lab water purification system is continuously able to meet the specifications for your water quality needs. Monitoring tools, such as resistivity monitors, which measure the ionic purity of water (a 18.2 MΩ·cm reading means that the product water is ion-free) and total oxidizable carbon (TOC) meters, which indicate the total amount of organic material present in water, are commonly used for this purpose. Depending on the application, both resistivity and TOC values may be required, so these criteria should be considered in your choice of water purification system.
Being able to trace and record water quality is also important, especially for labs working in regulated environments. Softwares and Web base solutions now provide the means to do with enhanced data management control, remote access capabilities to the system dashboard, and long-term electronic archiving. 
Some modern purification systems are equipped with digital touchscreens, which simplify water quality monitoring and make system operation more intuitive. Some more recent systems may also be remotely monitored and controlled, giving you greater assurance of your lab’s productivity when off-site.
User experience & service
To save time for everyone in the lab and gain efficiency, seek out a user-friendly system with an intuitive interface. Also, make sure cartridges are easy to change by anyone. Keeping your system in good condition with periodic preventive maintenance will ensure optimum operation throughout the equipment's lifetime. Check with your vendor to make sure that they have a professional service organization to meet your needs. Some water purification system suppliers also offer remote system monitoring and streamline consumable replacement, to optimize system uptime and lab productivity.
Consider certification and compliance
As part of the system selection process, you may also wish to investigate aspects concerning quality assurance. Look for systems that are produced in an ISO 9001- and ISO 14001- registered manufacturing site and which are delivered with the necessary quality documents, such as certificates attesting to conformity, quality, or calibration, for example, which will help you meet your Quality Management System requirements. Investing in a high-quality robust system ensures it will perform durably, delivering better value over time.
In addition, keep in mind that water purification systems must comply with regulations that govern efficiency and safety throughout the lifetime of the equipment: look for equipment with CE, UL, and FCC-certification.
The laboratory water produced by water purification systems is also subject to standards that define the quality required for specific applications, such as those published by ASTM® and ISO 3696 for laboratory applications, the Clinical and Laboratory Standards Institute (CLSI®) guidelines for clinical labs, or those provided by pharmacopoeial bodies. If the water used in your labs must comply with standards from these organizations, be sure to check this point, too. 
Strive for sustainability
It is true that the operation of any laboratory water purification system has a certain impact on the environment. However, manufacturers have made great progress in the development of sustainable solutions, and water systems on the market today reflect these changes in their reduced use of potable water, electricity, mercury and strong chemicals, and in the availability of purification cartridges with longer lifetimes. Some systems have become more compact, with smaller cartridges, to reduce plastic use. In addition, excessive packaging has often been reduced and documentation is usually provided as electronic files, rather than printed documents. In addition to the points above, check that your vendor's systems follow European Restriction of Hazardous Substances (RoHS) and Waste Electrical and Electronic Equipment (WEEE) directives.
Keeping your system in good condition with periodic preventive maintenance is also another way to reinforce sustainability and to ensure optimum operation throughout the equipment's lifetime. Check with your vendor to make sure that they have a professional service organization to meet your needs.
Don't forget design
By now you have determined where and when you need pure and ultrapure water, for which application(s), and in what volume and frequency. The actual design of the lab incorporates this information with other important criteria that should be discussed with your lab water purification vendor.
A larger lab may opt for a solution that involves distributing pure water through a loop, or implementing a floor-by-floor set up in response to volume or water purity needs, while smaller labs may prefer to use individual systems providing pure and ultrapure water on demand. In environments subject to regulatory requirements where full system control and complete traceability is needed, lab configuration may be determined by the need to install a number of point-of-use systems using compliant data record softwares.
Finally, don't forget that other stakeholders, such as members of building/facilities management or the purchasing department may need to be involved in the decision. Several criteria should be considered to guide the discussion with the water purification system vendor and select the solution.
Contact us if you would like support from a lab water expert in selecting a water system for your lab.
You can also try our Lab Water Selection Guide, an online tool that can help you identify and configure your optimal water purification system.
Related Products
Sorry, an unexpected error has occurred
Response not successful: Received status code 500