Positive Control siRNA
MISSION® Positive Control siRNA serve as an ideal complement to any gene silencing experiment. Positive controls are validated siRNA known to achieve high levels (>70%) of knockdown. A positive control should be used to optimize transfection, and if it fails to produce the expected phenotype, adjustments to experimental conditions are likely necessary.
Product Benefits
- Best-in-class, guaranteed gene silencing
- Knockdown efficiencies validated to be ≥75%
- Designed with the Rosetta algorithm*
*The proprietary Rosetta Inpharmatics siRNA Design algorithm is licensed in an exclusive partnership with MilliporeSigma. The Rosetta siRNA Design Algorithm utilizes Position-Specific Scoring Matrices and knowledge of the seed region to predict the most specific and effective sequences for your target genes. The algorithm’s rules were developed utilizing empirical data collected from gene silencing experiments carried out over three years.
Product Features
- Species: Human & Rat
- Quantities: 2 (10 nmol), 5 (25 nmol) & 10 (50 nmol) OD
- Purification: Desalt or HPLC
- Sequence Form: 21mer duplexes with dTdT overhangs
- Modifications: None (natural RNA bases only)
- Quality Control: 100% mass spectrometry*
- Format: Supplied dry in tubes
*Depending on manufacturing site, PAGE may be used to assess siRNA duplexes
If additional help is needed, please consult our technical services group at [email protected].
MISSION is a trademark of Merck KGaA, Darmstadt, Germany, and/or its affiliates. Label License.
Response not successful: Received status code 500