MilliporeSigma
HomeMedia FillsMedia Fills Test for Beverages
Media Fills Test for Beverages
Contamination of the filling lines in the beverage industry can cause a loss of millions of product units. It can also result in considerable reprocessing, disposal, and product-recall costs as well as damage to the brand reputation and consumer confidence. Media fill tests for beverages ensure that the filling line is not contaminated with spoilage bacteria, yeasts, or molds, during the production of low-acid aseptic beverages.
Glas with orange juice
LINDEN GRAIN MEDIUM FOR MEDIA FILL SIMULATIONS OF BEVERAGE BOTTLING AND ICE-CRUSHER INSTALLATIONS
Media fill test for beverages
Our ready-mixed granulated Linden Grain Medium is a high-quality alternative to powdered media and is ideal for the detection of yeasts, molds, and bacteria in beverages. It was successfully developed in cooperation with Coca-Cola HBC AG, Zug, Switzerland, for media fill simulations.
The nutrient content of the medium creates optimum growth conditions for fungi and bacteria. The pH of the medium can be adjusted to the filling line criteria (as low as pH 4.1), allowing the growth of fungi and bacteria that are adapted to grow under highly acidic conditions.
FEATURES AND BENEFITS OF LINDEN GRAIN MEDIUM
This high-quality medium is safe, easy to use, and ensures reliable results.
  • Ready-mixed and easily soluble
  • Granulated to minimize toxic and allergenic dust
  • Granules reduce on-site contamination with media dust
  • Meets highest performance standards in production and QC as described in major company standards (ISO 11133)
  • Available in 5 kg and cost-saving 25 kg sizes
We also offer halal & kosher-certified Linden Grain Medium
  • Identical in composition and performance
  • Halal certified acc. to JAKIM standards
  • Certified kosher by KF Kosher
  • Available in powder format
SUPERIOR POWDERED MEDIA FOR MEDIA FILL TESTS
Our Linden Grain Medium is typically prepared in large volumes of more than 5,000 liters for a single media fill test – no need to source, buy, mix, and quality control individual raw materials.
The unique granulated format of Linden Grain Medium ensures:
  • Excellent solubility and free-flowing properties
  • No component separation and clumping, even under warm or humid conditions
  • Homogenous distribution of non-animal-based ingredients
  • Less dust formation. Reduces cleaning efforts and limits the spread of media components that may be hazardous or trigger allergic reactions
SUITABILITY OF LINDEN GRAIN MEDIUM FOR ICE-CRUSHER TESTING
Linden Grain Medium possesses properties that make it suitable for microbial testing of ice-crusher installations.
  • Stable after freezing and thawing (color, pH)
  • Performance after freezing comparable to freshly prepared medium
CONSISTENT HIGH-QUALITY MEDIA FILL IN ASEPTIC ENVIRONMENTS
Only carefully selected raw materials are used to manufacture media fills for the beverage industry. Our quality-control procedures include testing the granulated Linden Grain Medium with typical beverage spoilage strains. These results indicate not only excellent performance (see Table 1) but also a level of batch-to-batch consistency that is difficult to achieve when producing Linden Grain Medium by yourself.
Coca-Cola HBC AG, one of the world’s largest cola drinks bottlers, with operations in 28 countries, successfully applies Linden Grain Medium in its aseptic operations.
Product No.Description
Linden Grain Medium
1.03790Linden Grain Medium suitable for sterility testing, for yeasts, for molds, for bacteria
1.02747Linden Grain Medium Halal (Base) suitable for sterility testing, for yeasts, for molds, for bacteria
Chemicals for pH adjustment of Linden Grain Medium
1.00314Hydrochloric acid fuming, 37%, EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, NF
1.00804L(+)-Tartaric acid for analysis EMSURE® ACS, ISO, Reag. Ph Eur
Table 1.Typical spoilage microorganisms of low-acid beverages: Growth in Millipore® Linden Grain Medium (B. subtilis used as negative growth control).
COMPREHENSIVE SOLUTIONS
Typically, a media fill simulation is accompanied by viable and non-viable air monitoring and surface monitoring of the production environment.
Viable Air Monitoring
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Surface Monitoring
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Rapid Hygiene Monitoring
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Related Webinar