Merck
  • Occurrence and fate of PPCPs and correlations with water quality parameters in urban riverine waters of the Pearl River Delta, South China.
Occurrence and fate of PPCPs and correlations with water quality parameters in urban riverine waters of the Pearl River Delta, South China.
Environmental science and pollution research international (2013-04-24)
Xin Yang, Feng Chen, Fangang Meng, Yuanyu Xie, Hui Chen, Kyana Young, Wangxing Luo, Tingjin Ye, Wenjie Fu
ZUSAMMENFASSUNG
The occurrence and fate of eight PPCPs was studied in river waters from upstream to downstream of the three rivers in the Pearl River Delta, China. The correlations of PPCP levels and water quality parameters were also investigated. The analytes of the highest concentrations were caffeine, acetaminophen, and ciprofloxacin. Carbamazepine and erythromycin-H2O were detected at the lowest concentrations. The highest concentrations of PPCPs were found in the Shijing River, with 865 ng/L caffeine, 339 ng/L acetaminophen, and 304 ng/L ciprofloxacin. In general, the levels of PPCPs in the Zhujiang River were higher at sites where the metropolitan city Guangzhou is located and decreased from the epicenter along the river. Low levels of PPCPs were generally found in the Beijiang River. Positive correlations were found between PPCP levels, total nitrogen, ammonium nitrogen, and cumulative fluorescence excitation-emission matrix (EEM) volume. Among the four PPCPs evaluated (caffeine, acetaminophen, ciprofloxacin, and sulfamethoxazole), caffeine had the best correlations with the correlation coefficients ranging from 0.62 to 0.98. The prediction of PPCP concentrations at specified locations can be substantially simplified.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Koffein, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Koffein, powder, ReagentPlus®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Trimethoprim, ≥98.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Koffein, anhydrous, 99%, FCC, FG
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Carbamazepin, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Schmelzpunktstandard 235-237°C, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Coffein Schmelzpunktstandard, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ciprofloxacin HCl, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Koffein -Lösung, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
N,N-Diethyl-3-Methylbenzamid, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Koffein, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Trimethoprim, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Koffein, Sigma Reference Standard, vial of 250 mg
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Erythromycin-Standard -Lösung, 1 mg/mL in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
DEET, PESTANAL®, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Carbamazepin, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Koffein, meets USP testing specifications, anhydrous
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Koffein -Lösung, analytical standard, 1.0 mg/mL in methanol
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Carbamazepine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Mettler Toledo Calibration Substance ME 18872, Caffeine, traceable to primary standards (LGC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Trimethoprim, VETRANAL®, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Koffein, anhydrous, tested according to Ph. Eur.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Ciprofloxacin-Hydrochlorid, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Koffein, BioXtra
Preise und Verfügbarkeit sind derzeit nicht verfügbar.