Merck
Genotoxic and cell-transforming effects of titanium dioxide nanoparticles.
Environmental research (2014-12-03)
Eşref Demir, Hakan Akça, Fatma Turna, Sezgin Aksakal, Durmuş Burgucu, Bülent Kaya, Onur Tokgün, Gerard Vales, Amadeu Creus, Ricard Marcos
ZUSAMMENFASSUNG
The in vitro genotoxic and the soft-agar anchorage independent cell transformation ability of titanium dioxide nanoparticles (nano-TiO2) and its microparticulated form has been evaluated in human embryonic kidney (HEK293) and in mouse embryonic fibroblast (NIH/3T3) cells. Nano-TiO2 of two different sizes (21 and 50 nm) were used in this study. The comet assay, with and without the use of FPG enzyme, the micronucleus assay and the soft-agar colony assay were used. For both the comet assay and the frequency of micronuclei a statistically significant induction of DNA damage, was observed at the highest dose tested (1000 µg/mL). No oxidative DNA damage induction was observed when the comet assay was complemented with the use of FPG enzyme. Furthermore, long-term exposure to nano-TiO2 has also proved to induce cell-transformation promoting cell-anchorage independent growth in soft-agar. Results were similar for the two nano-TiO2 sizes. Negative results were obtained when the microparticulated form of TiO2 was tested, indicating the existence of important differences between the microparticulated and nanoparticulated forms. As a conclusion it should be indicated that the observed genotoxic/tranforming effects were only detected at the higher dose tested (1000 µg/mL) what play down the real risk of environmental exposures to this nanomaterial.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Titan(IV)-oxid, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, ReagentPlus®, ≥99%
Sigma-Aldrich
Titan(IV)-oxid, Anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Rutil, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Rutil, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Mischung aus Rutil und Anatas, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titan, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titan, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titan, powder, <45 μm avg. part. size, 99.98% trace metals basis
Sigma-Aldrich
Titan, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Mischung aus Rutil und Anatas, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Rutil, ≥99.98% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Rutil, 99.995% trace metals basis
Titan, mesh, 100x100mm, nominal aperture 0.19mm, wire diameter 0.23mm, 60x60 wires/inch, open area 20%, twill weave
Sigma-Aldrich
Titan(IV)-oxid, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Sigma-Aldrich
Titan, sponge, 1-20 mm, 99.5% trace metals basis
Sigma-Aldrich
Titan, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titan, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.995% trace metals basis
Titan, mesh, 100x100mm, nominal aperture 4.3mm, wire diameter 1.5mm, 4.4x4.4 wires/inch, open area 94%, platinized diamond mesh
Sigma-Aldrich
Titan, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Titan, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Titan, ≤10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titan, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Rutil, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Titan, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, nanowires, diam. × L ~10 nm × 10 μm
Sigma-Aldrich
Titan, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, nanowires, diam. × L ~100 nm × 10 μm