Merck
  • Fractionation of complex lipid mixtures by hydroxyapatite chromatography for lipidomic purposes.
Fractionation of complex lipid mixtures by hydroxyapatite chromatography for lipidomic purposes.
Journal of chromatography. A (2014-08-26)
Gabriella Pinto, Simonetta Caira, Gianfranco Mamone, Pasquale Ferranti, Francesco Addeo, Gianluca Picariello
ZUSAMMENFASSUNG
The comprehensive analysis of natural lipid mixtures is often challenging due to the simultaneous occurrence of functionally and structurally heterogeneous compounds. Modern analytical approaches in system-wide lipidomics essentially rely on mass spectrometry (MS). The overabundant amount of triacylglycerols (TAGs) in most samples hinders the direct detection of phospholipids (PLs) or other low-abundance lipids; therefore, a fractionation step is most often required to reduce sample complexity prior to MS analysis. In this work, we explore the use of hydroxyapatite (HAP) as a chromatographic stationary phase (gel HAP) or chemo-affinity sorbent material (ceramic HAP) to selectively enrich PLs and polar lipids from chicken egg yolk and buttermilk. Due to the affinity of phosphate-containing compounds for HAP, both in-column and in-batch HAP-based chromatography were effective to deplete TAGs before releasing PLs with an eluting ternary system containing sodium phosphate as the displacing agent. Buttermilk gangliosides and PLs co-eluted, indicating that HAP also exhibits a high affinity for sialylated glycolipids and that polar lipids are retained through a combined mechanism of chemo-affinity and hydrogen bonding. To the best of our knowledge, this is the first report in which HAP is proposed as an alternative stationary phase for separating both the PLs and sialylated glycolipids from TAGs in complex lipidomes. The HAP-based chromatography has potential to be improved for the separation of the polar lipid classes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
2,5-Dihydroxybenzoesäure, 98%
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%