Merck
  • Detailed characterisation of circulatory nitric oxide and free radical indices--is there evidence for abnormal cardiovascular homeostasis in young women with polycystic ovary syndrome?
Detailed characterisation of circulatory nitric oxide and free radical indices--is there evidence for abnormal cardiovascular homeostasis in young women with polycystic ovary syndrome?
BJOG : an international journal of obstetrics and gynaecology (2014-05-13)
G R Willis, M Udiawar, W D Evans, H L Blundell, P E James, D A Rees
ZUSAMMENFASSUNG
To assess circulating biochemical indices of endothelial function and nitro-oxidative stress in women with polycystic ovary syndrome (PCOS). Case-control study. Seventeen women with PCOS and eighteen age- and body mass index-matched healthy volunteers. Nitric oxide (NO) metabolite levels were assessed by chemiluminescence. Electron paramagnetic resonance spectroscopy with spin trapping was used to assess oxidative stress ex vivo and in vitro. Antioxidant capacity was measured using oxygen radical absorbance. Biochemical indices of endothelial function, including NO metabolites, lipid-derived radicals and antioxidant capacity. Plasma NO metabolites were similar in the two groups (nitrite: 257±116 nmol/l [PCOS], 261±135 nmol/l [controls] P=0.93; nitrate: 27±7 μmol/l [PCOS], 26±6 μmol/l [controls] P=0.89). Alkoxyl free radicals (lipid-derived) were detected as the dominant species, but levels were not different between women with PCOS and controls whether measured directly ex vivo (median 7.2 [range 0.17-16.73]e6 arbitrary units [a.u.] and 7.2 [1.7-11.9]e6 a.u., respectively, P=0.57) or when stimulated in vitro to test radical generation capacity (1.23 [0.3-5.62]e7 a.u. and 1.1 [0.48-15.7]e7 a.u. respectively, P=0.71). In regression analysis, visceral fat area was independently associated with in vitro oxidative potential (β=0.6, P=0.002). Total plasma antioxidant capacity (94±30% [PCOS], 79±24% [controls], P=0.09) and plasma hydroperoxides (7.5±4 μmol/l [PCOS], 6.7±5 μmol/l [controls], P=0.21) were not different between groups. However, lipophilic antioxidant capacity was lower in women with PCOS compared with controls (92±32 and 125±48%, respectively, P=0.02). Young overweight women with PCOS display a reduced lipophilic antioxidant capacity compared with healthy volunteers, but no change in circulating free radicals or nitro-oxidative stress.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Fluorescein, suitable for fluorescence, free acid
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%