Merck
  • Differential expression of miRNAs and their target mRNAs in endometria prior to maternal recognition of pregnancy associates with endometrial receptivity for in vivo- and in vitro-produced bovine embryos.
Differential expression of miRNAs and their target mRNAs in endometria prior to maternal recognition of pregnancy associates with endometrial receptivity for in vivo- and in vitro-produced bovine embryos.
Biology of reproduction (2014-09-26)
Siriluck Ponsuksili, Dawit Tesfaye, Karl Schellander, Michael Hoelker, Frieder Hadlich, Manfred Schwerin, Klaus Wimmers
ZUSAMMENFASSUNG
Endometrial receptivity is a prerequisite for successful embryo implantation and pregnancy. Receptivity involves complex processes promoted by many transcripts that are key components of molecular pathways that depend on ovarian hormones and that contribute to shaping structural, metabolic, and communication properties of endometrial cells toward reception of embryos. MicroRNAs (miRNAs) are important regulators of the expression of these transcripts encoding effector molecules. We acquired miRNA and mRNA signatures, miRNA-mRNA pairs, and regulatory networks linked with the emergence and maintenance of postimplantation pregnancy. Endometrial tissue samples were obtained at Days 3 and 7 of the estrous cycle of cows that did or did not become pregnant after transfer of either in vivo-produced (IVV) or in vitro-produced (IVT) embryos in the next cycle following the biopsy. We report a list of endometrial miRNAs that were differentially expressed between Day 3 and Day 7 of the bovine estrous cycle (including miR-1290, miR-3437, miR-1246, miR-486, miR-3107, and miR-382), that differed with high or low endometrial receptivity (miR-3902-3p, miR-1825, miR-H14-3p, miR-885-3p, miR-504-3p, and miR-186), or that differed among the IVT and IVV transfers (miR-449a/b/c, miR-138, miR-874, miR-4342, miR-2231, and miR-2751). Moreover, mRNA transcripts were also analyzed, and pairs of negatively correlated miRNAs and mRNAs were predicted in silico. The miRNA-mRNA target pairs had roles in response to hormonal stimuli and oxidative stress, chromatin organization, miRNA-mediated epigenetic histone changes, cell proliferation, p53 signaling, and apoptosis. Overall, we identified significant miRNAs, miRNA-mRNA pairs, and functional networks that are associated with the state of pregnancy at Day 28 as a parameter of endometrial receptivity and that are affected by estrous cycle and embryo culture systems.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Phenol -Lösung, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Phenol -Lösung, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Verflüssigtes Phenol, ≥89.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Supelco
Phenol -Lösung, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol -Lösung, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)