Merck
  • Interindividual differences in response to treatment with butyrate-producing Butyricicoccus pullicaecorum 25-3T studied in an in vitro gut model.
Interindividual differences in response to treatment with butyrate-producing Butyricicoccus pullicaecorum 25-3T studied in an in vitro gut model.
FEMS microbiology ecology (2015-05-23)
Annelies Geirnaert, Jun Wang, Magali Tinck, Alix Steyaert, Pieter Van den Abbeele, Venessa Eeckhaut, Ramiro Vilchez-Vargas, Gwen Falony, Debby Laukens, Martine De Vos, Filip Van Immerseel, Jeroen Raes, Nico Boon, Tom Van de Wiele
ZUSAMMENFASSUNG
Butyrate-producing bacteria are promising probiotic candidates to target microbial dysbiosis in gastrointestinal disorders like inflammatory bowel diseases. Butyricicoccus pullicaecorum 25-3(T), a butyrate-producing clostridial cluster IV strain, is such a candidate. Little is known about its abundance in the colon microbiota and its butyrogenic properties. We used the M-SHIME(®), an in vitro simulator for the human intestinal microbial ecosystem, to study the effect of supplementing a single dose of B. pullicaecorum 25-3(T) on lumen- and mucus-associated microbiota of eight individuals. Butyricicoccus pullicaecorum was more abundant in mucus-associated microbiota compared with lumen microbiota. Supplementation with a single dose of B. pullicaecorum 25-3(T) resulted in a temporary increase in B. pullicaecorum bacteria in lumen compartment of all individuals. In two cases, the responders, an increased butyrate production was observed as compared with the control. 16S rRNA gene amplicon sequencing revealed the microbiota of responders to be different as compared to non-responder microbiota. We can conclude that B. pullicaecorum 25-3(T) is a mucus-associated bacterium whose potency to stimulate butyrate production is characterized by a large interindividual variability in terms of composition of the receiving microbial community.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Cystein, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Cystein, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Cystein, BioUltra, ≥98.5% (RT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumbicarbonat, BioXtra, 99.5-100.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
L-Cystein
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Cystein, ≥97%, FG
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Cystein, Wacker Chemie AG, ≥98.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumbicarbonat-12C, 13C-depleted, 99.9 atom % 12C
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumchlorid -Lösung, 0.1 M
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~0.025 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.