Merck
  • Radical Cation/Radical Reactions: A Fourier Transform Ion Cyclotron Resonance Study of Allyl Radical Reacting with Aromatic Radical Cations.
Radical Cation/Radical Reactions: A Fourier Transform Ion Cyclotron Resonance Study of Allyl Radical Reacting with Aromatic Radical Cations.
International journal of mass spectrometry (2010-04-20)
Amber L Russell, Henry W Rohrs, David Read, Daryl E Giblin, Peter P Gaspar, Michael L Gross
RESUMO
A method for the study of reactions of open-shell neutrals (radicals) and radical cations is described. Pyrolysis (25-1500 degrees C) of thermally labile compounds, such as, 1,5-hexadiene via a Chen nozzle yields a seeded beam of reactive species in helium. The pyrolysis products are then analyzed by electron ionization (EI) or reacted with stored ions. Electron ionization of the pyrolysis products of 1,5-hexadiene shows that both the allyl radical and allene are generated. Reactions of benzene radical cations and the pyrolysis products of 1,5-hexadiene result in carbon-carbon bond formation. Those reactions of allyl radical with the benzene radical cation yield the C(7)H(7) (+) ion of m/z 91, permitting an unusual entry into arenium ions. The reaction of allene with benzene radical cation in contrast yields C(9)H(10) (+). and C(9)H(9) (+).
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
1,5-Hexadiene, 97%
Preço e disponibilidade não estão disponíveis no momento.