Merck
Spermine and arcaine block and permeate N-methyl-D-aspartate receptor channels.
Biophysical journal (1999-06-04)
R C Araneda, J Y Lan, X Zheng, R S Zukin, M V Bennett
RESUMO
Polyamines such as spermine are thought to be endogenous regulators of NMDA (N-methyl-D-aspartate)-type glutamate receptors. Polyamine block of NMDA receptors was studied in excised outside-out patches from rat hippocampal neurons and Xenopus oocytes expressing recombinant receptors. Extracellular spermine and arcaine reduced NMDA single-channel conductance in a voltage-dependent manner, with partial relief of block evident at large inside negative membrane potentials. Reducing extracellular Na+ concentration increased the apparent affinities for spermine and arcaine, indicating strong interaction between spermine and permeant ions. Internal spermine also blocked NMDA channels in a voltage-dependent manner, with relief of block evident at large inside positive potentials. The Woodhull model of channel block by an impermeant ion adequately described the actions of external spermine from -60 to +60 mV, but failed for more negative potentials. Eyring rate theory for a permeable blocker with two barriers and one binding site adequately described the voltage-dependent block and relief from block by both external and internal spermine over the range of -120 to +60 mV. These findings indicate that polyamines block and permeate neuronal NMDA receptor channels from the extracellular and intracellular sides, although sensitivity to internal spermine is probably too low to be physiologically relevant.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Arcaine sulfate salt
Preço e disponibilidade não estão disponíveis no momento.