Virus Cultivation
Primary, Human Diploid, and Continuous Culture Cell Lines for Virus Isolation and Proliferation and Virus-based Vaccines
The advent of cell culture techniques has fundamentally changed virus isolation and proliferation in the lab setting. Cell-based production systems offer a convenient and cost-effective approach for the isolation, detection, and identification of viruses. Greater process control contributes to a more reliable and well-characterized product, with faster and shorter production cycles than that of animal- and egg-based production systems.
Cell-based production systems for virus culture and vaccine production are important for:
- Virus detection/identification: Cell cultures provide a suitable environment for detection and identification of many human viral pathogens, affording important microscopic examinations for evidence of viral proliferation. Accurate identification of virus is important to ensure timely and appropriate treatments, and can facilitate the detection of mixed viral infections.
- Host-pathogen interaction research: Innovations in cell biology have allowed deeper and more complex insights into host-pathogen interactions for the study of pathogenesis. In vitro cell culture systems can facilitate experimental access for investigation of the mode and etiological factors of viral infection.
- Viral structure and replication: Genetic material and replication methods vary considerably among different types of viruses. Cell culture systems can facilitate virus growth and elucidate development and interactions with host cells at every stage of replication.
- Vaccine production: Cell-based vaccine production systems offer a flexible and cost-effective approach for meeting vaccine output needs. Manufacturers can supply vaccines more quickly and in greater quantities to alleviate vaccine supply shortages during outbreaks when traditional egg-based production systems may fall short. Virus-based vaccines produced in mammalian cells may also offer better protection against viral infections, as they more closely replicate viruses in circulation than vaccines produced in chicken eggs.
Cell culture systems used for virus propagation may employ primary cells, semi-continuous cell lines, and continuous cell lines:
| Primary cell lines | Human diploid cells (Semi-continuous cell lines) | Continuous cell lines |
|---|---|---|
|
|
|
| Cell Line | Origin | Infectious Agents |
|---|---|---|
| A549 | human lung carcinoma | Adenovirus,1 HSV,2 influenza,3 measles,4 mumps,4 parainfluenza,5 poliovirus,6 respiratory syncytial virus (RSV),6 rotavirus,7 Varicella zoster virus (VZV),8 metapneumovirus (MPV)9 |
| BHK 21 (clone 13) | Syrian hamster kidney | Human adenovirus D,10 reovirus 3,11 vesicular stomatitis virus (Indiana strain),12 Dengue,13 influenza,13 rabies,13 foot and mouth,14 rubella,15 |
| CV-1 | African green monkey kidney fibroblast | RSV,16, measles,16, HSV,17, VZV,18 |
| HeLa | human cervix adenocarcinoma | Poliovirus type I,19 adenovirus type 3,20 CMV,21 echovirus,22 HSV23 poliovirus,24 rhinovirus,25 vesicular stomatitis (Indiana Strain) virus,26 VZV27 |
| LLCMK2 | Rhesus monkey kidney | Poliovirus type 1,17 enterovirus,28 rhinovirus,29 poxvirus groups30 |
| McCoy | Mouse fibroblast | HSV32 |
| MDCK | Madin-Darby canine kidney | Influenza A,31 influenza B, some types of adenovirus,33 reoviruses34 |
| MRC-5 | human fetal lung | CMV,35 HSV,36 adenovirus,37 influenza,38 mumps,39 echovirus,40 poliovirus,41 rhinovirus,42 RSV,43 VZV,27 |
| NCI-H292 | Human lung, mucoepidermoid carcinoma | Vaccinia virus,44 HSV,45 adenovirus,46 measles virus,47 reoviruses,48 BK polyomavirus,49 RSV,50 some strains of influenza A,51 most enteroviruses,52 and rhinoviruses53 |
| Vero | African green monkey kidney | Coxsackie B,54 HSV,55 measles,56 mumps,57 poliovirus type 3,58 rotavirus,59 rubella,60 |
| Vero76 | African green monkey kidney | Coxsackie B,61 HSV,62 West Nile virus,63 |
| Wi 38 | Human fetal lung | Adenovirus,64 CMV,65 echovirus,66 HSV,67 mumps,68 influenza,69 rhinovirus,70 RSV,71 VZV,72 |
References
1.
Hodzic J, Sie D, Vermeulen A, van Beusechem VW. 2017. Functional Screening Identifies Human miRNAs that Modulate Adenovirus Propagation in Prostate Cancer Cells. Human Gene Therapy. 28(9):766-780. https://doi.org/10.1089/hum.2016.143
2.
Wang Y, Zhou B, Lu J, Chen Q, Ti H, Huang W, Li J, Yang Z, Jiang Z, Wang X. 2017. Inhibition of influenza virus via a sesquiterpene fraction isolated from Laggera pterodonta by targeting the NF-?B and p38 pathways. BMC Complement Altern Med. 17(1): https://doi.org/10.1186/s12906-016-1528-8
3.
Holzberg M, Boergeling Y, Schräder T, Ludwig S, Ehrhardt C. Vemurafenib Limits Influenza A Virus Propagation by Targeting Multiple Signaling Pathways. Front. Microbiol.. 8 https://doi.org/10.3389/fmicb.2017.02426
4.
Franz S, Rennert P, Woznik M, Grützke J, Lüdde A, Arriero Pais EM, Finsterbusch T, Geyer H, Mankertz A, Friedrich N. 2017. Mumps Virus SH Protein Inhibits NF-?B Activation by Interacting with Tumor Necrosis Factor Receptor 1, Interleukin-1 Receptor 1, and Toll-Like Receptor 3 Complexes. J Virol. 91(18): https://doi.org/10.1128/jvi.01037-17
5.
Dirr L, El-Deeb IM, Chavas LMG, Guillon P, Itzstein Mv. 2017. The impact of the butterfly effect on human parainfluenza virus haemagglutinin-neuraminidase inhibitor design. Sci Rep. 7(1): https://doi.org/10.1038/s41598-017-04656-y
6.
McCaskill JL, Ressel S, Alber A, Redford J, Power UF, Schwarze J, Dutia BM, Buck AH. 2017. Broad-Spectrum Inhibition of Respiratory Virus Infection by MicroRNA Mimics Targeting p38 MAPK Signaling. Molecular Therapy - Nucleic Acids. 7256-266. https://doi.org/10.1016/j.omtn.2017.03.008
7.
Mackowiak M, Leifels M, Hamza IA, Jurzik L, Wingender J. 2018. Distribution of Escherichia coli, coliphages and enteric viruses in water, epilithic biofilms and sediments of an urban river in Germany. Science of The Total Environment. 626650-659. https://doi.org/10.1016/j.scitotenv.2018.01.114
8.
Aiba N, Shiraki A, Yajima M, Oyama Y, Yoshida Y, Ohno A, Yamada H, Takemoto M, Daikoku T, Shiraki K. 2017. Interaction of Immunoglobulin with Cytomegalovirus-Infected Cells. Viral Immunology. 30(7):500-507. https://doi.org/10.1089/vim.2016.0151
9.
Li Y, Lund C, Nervik I, Loevenich S, Døllner H, Anthonsen MW, Johnsen IB. 2018. Characterization of signaling pathways regulating the expression of pro-inflammatory long form thymic stromal lymphopoietin upon human metapneumovirus infection. Sci Rep. 8(1): https://doi.org/10.1038/s41598-018-19225-0
10.
Okeke M, Okoli A, Diaz D, Offor C, Oludotun T, Tryland M, Bøhn T, Moens U. Hazard Characterization of Modified Vaccinia Virus Ankara Vector: What Are the Knowledge Gaps?. Viruses. 9(11):318. https://doi.org/10.3390/v9110318
11.
Pudupakam RS, Raghunath S, Pudupakam M, Daggupati S. 2017. Genetic characterization of the non-structural protein-3 gene of bluetongue virus serotype-2 isolate from India. Vet World. 10(3):348-352. https://doi.org/10.14202/vetworld.2017.348-352
12.
Kleinlützum D, Hanauer JDS, Muik A, Hanschmann K, Kays S, Ayala-Breton C, Peng K, Mühlebach MD, Abel T, Buchholz CJ. Enhancing the Oncolytic Activity of CD133-Targeted Measles Virus: Receptor Extension or Chimerism with Vesicular Stomatitis Virus Are Most Effective. Front. Oncol.. 7 https://doi.org/10.3389/fonc.2017.00127
13.
Nikolay A, Castilho LR, Reichl U, Genzel Y. 2018. Propagation of Brazilian Zika virus strains in static and suspension cultures using Vero and BHK cells. Vaccine. 36(22):3140-3145. https://doi.org/10.1016/j.vaccine.2017.03.018
14.
Kamal SA, El-Rahman Hassan RA. Advanced Virological And Clinicopathological Studies On Cattle Suffering From Foot And Mouth Disease Virus. JI. 1(1):33-47. https://doi.org/10.14302/issn.2577-137x.ji-17-1736
15.
Prescott J, Feldmann H, Safronetz D. 2017. Amending Koch's postulates for viral disease: When ?growth in pure culture? leads to a loss of virulence. Antiviral Research. 1371-5. https://doi.org/10.1016/j.antiviral.2016.11.002
16.
Hampton CM, Strauss JD, Ke Z, Dillard RS, Hammonds JE, Alonas E, Desai TM, Marin M, Storms RE, Leon F, et al. 2017. Correlated fluorescence microscopy and cryo-electron tomography of virus-infected or transfected mammalian cells. Nat Protoc. 12(1):150-167. https://doi.org/10.1038/nprot.2016.168
17.
Dotti S, Lombardo T, Villa R, Cacciamali A, Zanotti C, Andreani NA, Cinotti S, Ferrari M. Transformation and Tumorigenicity Testing of Simian Cell Lines and Evaluation of Poliovirus Replication. PLoS ONE. 12(1):e0169391. https://doi.org/10.1371/journal.pone.0169391
18.
Yang K, Dang X, Baines JD. 2017. A Domain of Herpes Simplex Virus pUL33 Required To Release Monomeric Viral Genomes from Cleaved Concatemeric DNA. J Virol. 91(20): https://doi.org/10.1128/jvi.00854-17
19.
Gromeier M, Nair SK. 2018. Recombinant Poliovirus for Cancer Immunotherapy. Annu. Rev. Med.. 69(1):289-299. https://doi.org/10.1146/annurev-med-050715-104655
20.
Weigert M, Binks A, Dowson S, Leung EYL, Athineos D, Yu X, Mullin M, Walton JB, Orange C, Ennis D, et al. 2017. RIPK3 promotes adenovirus type 5 activity. Cell Death Dis. 8(12): https://doi.org/10.1038/s41419-017-0110-8
21.
Khadivjam B, Stegen C, Hogue-Racine M, El Bilali N, Döhner K, Sodeik B, Lippé R. 2017. The ATP-Dependent RNA Helicase DDX3X Modulates Herpes Simplex Virus 1 Gene Expression. J Virol. 91(8): https://doi.org/10.1128/jvi.02411-16
22.
Aguilera ER, Erickson AK, Jesudhasan PR, Robinson CM, Pfeiffer JK. 2017. Plaques Formed by Mutagenized Viral Populations Have Elevated Coinfection Frequencies. mBio. 8(2): https://doi.org/10.1128/mbio.02020-16
23.
Ibáñez FJ, Farías MA, Retamal-Díaz A, Espinoza JA, Kalergis AM, González PA. Pharmacological Induction of Heme Oxygenase-1 Impairs Nuclear Accumulation of Herpes Simplex Virus Capsids upon Infection. Front. Microbiol.. 8 https://doi.org/10.3389/fmicb.2017.02108
24.
Acevedo A, Woodman A, Arnold JJ, Yeh MT, Evans D, Cameron CE, Andino R. Genetic recombination of poliovirus facilitates subversion of host barriers to infection. https://doi.org/10.1101/273060
25.
Ganjian H, Zietz C, Mechtcheriakova D, Blaas D, Fuchs R. ICAM-1 Binding Rhinoviruses Enter HeLa Cells via Multiple Pathways and Travel to Distinct Intracellular Compartments for Uncoating. Viruses. 9(4):68. https://doi.org/10.3390/v9040068
26.
Pan W, Song D, He W, Lu H, Lan Y, Li H, Gao F, Zhao K. 2017. EIF3i affects vesicular stomatitis virus growth by interacting with matrix protein. Veterinary Microbiology. 21259-66. https://doi.org/10.1016/j.vetmic.2017.10.021
27.
Sadaoka T, Schwartz CL, Rajbhandari L, Venkatesan A, Cohen JI. 2017. Human Embryonic Stem Cell-Derived Neurons Are Highly Permissive for Varicella-Zoster Virus Lytic Infection. J Virol. 92(1): https://doi.org/10.1128/jvi.01108-17
28.
Betancourt WQ, Abd-Elmaksoud S, Gerba CP. 2018. Efficiency of Reovirus Concentration from Water with Positively Charged Filters. Food Environ Virol. 10(2):209-211. https://doi.org/10.1007/s12560-017-9332-2
29.
Sato K, Watanabe O, Ohmiya S, Chiba F, Suzuki A, Okamoto M, Younghuang J, Hata A, Nonaka H, Kitaoka S, et al. 2017. Efficient isolation of human metapneumovirus using MNT-1, a human malignant melanoma cell line with early and distinct cytopathic effects. Microbiol Immunol. 61(11):497-506. https://doi.org/10.1111/1348-0421.12542
30.
Parker S, Crump R, Hartzler H, Buller R. Evaluation of Taterapox Virus in Small Animals. Viruses. 9(8):203. https://doi.org/10.3390/v9080203
31.
Petyaev IM, Zigangirova NA, Morgunova EY, Kyle NH, Fedina ED, Bashmakov YK. 2017. Resveratrol Inhibits Propagation ofChlamydia trachomatisin McCoy Cells. BioMed Research International. 20171-7. https://doi.org/10.1155/2017/4064071
32.
Danaher RJ, Fouts DE, Chan AP, Choi Y, DePew J, McCorrison JM, Nelson KE, Wang C, Miller CS. 2017. HSV-1 clinical isolates with unique in vivo and in vitro phenotypes and insight into genomic differences. J. Neurovirol.. 23(2):171-185. https://doi.org/10.1007/s13365-016-0485-9
33.
Glatthaar-Saalmüller B, Mair KH, Saalmüller A. 2017. Antiviral activity of aspirin against RNA viruses of the respiratory tract-an in vitro study. Influenza Other Respi Viruses. 11(1):85-92. https://doi.org/10.1111/irv.12421
34.
Shah Mahmud R, Müller C, Romanova Y, Mostafa A, Ulyanova V, Pleschka S, Ilinskaya O. 2017. Ribonuclease fromBacillusActs as an Antiviral Agent against Negative- and Positive-Sense Single Stranded Human Respiratory RNA Viruses. BioMed Research International. 20171-11. https://doi.org/10.1155/2017/5279065
35.
Hobom U, Brune W, Messerle M, Hahn G, Koszinowski UH. 2000. Fast Screening Procedures for Random Transposon Libraries of Cloned Herpesvirus Genomes: Mutational Analysis of Human Cytomegalovirus Envelope Glycoprotein Genes. J. Virol.. 74(17):7720-7729. https://doi.org/10.1128/jvi.74.17.7720-7729.2000
36.
Didcock L, Young DF, Goodbourn S, Randall RE. 1999. Sendai Virus and Simian Virus 5 Block Activation of Interferon-Responsive Genes: Importance for Virus Pathogenesis. J. Virol.. 73(4):3125-3133. https://doi.org/10.1128/jvi.73.4.3125-3133.1999
37.
Gugala Z, Olmsted-Davis EA, Gannon FH, Lindsey RW, Davis AR. 2003. Osteoinduction by ex vivo adenovirus-mediated BMP2 delivery is independent of cell type. Gene Ther. 10(16):1289-1296. https://doi.org/10.1038/sj.gt.3302006
38.
Cheng P, Ng L, Chiang L, Lin C. 2006. ANTIVIRAL EFFECTS OF SAIKOSAPONINS ON HUMAN CORONAVIRUS 229E IN VITRO. Clin Exp Pharmacol Physiol. 33(7):612-616. https://doi.org/10.1111/j.1440-1681.2006.04415.x
39.
Wilson MR, Suan D, Duggins A, Schubert RD, Khan LM, Sample HA, Zorn KC, Rodrigues Hoffman A, Blick A, Shingde M, et al. 2017. A novel cause of chronic viral meningoencephalitis: Cache Valley virus. Ann Neurol.. 82(1):105-114. https://doi.org/10.1002/ana.24982
40.
Atieh T, El Ayoubi MD, Aubry F, Priet S, de Lamballerie X, Nougairède A. Haiku: New paradigm for the reverse genetics of emerging RNA viruses. PLoS ONE. 13(2):e0193069. https://doi.org/10.1371/journal.pone.0193069
41.
Lewandowska DW, Zagordi O, Geissberger F, Kufner V, Schmutz S, Böni J, Metzner KJ, Trkola A, Huber M. 2017. Optimization and validation of sample preparation for metagenomic sequencing of viruses in clinical samples. Microbiome. 5(1): https://doi.org/10.1186/s40168-017-0317-z
42.
Jenny RA, Hirst C, Lim SM, Goulburn AL, Micallef SJ, Labonne T, Kicic A, Ling K, Stick SM, Ng ES, et al. 2015. Productive Infection of Human Embryonic Stem Cell-Derived NKX2.1+Respiratory Progenitors With Human Rhinovirus. 4(6):603-614. https://doi.org/10.5966/sctm.2014-0274
43.
Jerome KR. Lennette's Laboratory Diagnosis of Viral Infections. https://doi.org/10.3109/9781420084962
44.
45.
Mu J, Hirayama M, Sato Y, Morimoto K, Hori K. A Novel High-Mannose Specific Lectin from the Green Alga Halimeda renschii Exhibits a Potent Anti-Influenza Virus Activity through High-Affinity Binding to the Viral Hemagglutinin. Marine Drugs. 15(8):255. https://doi.org/10.3390/md15080255
46.
Costello MJ, Smernoff NT, Yungbluth M. 1993. Laboratory Diagnosis of Viral Respiratory Tract Infections. Lab Med. 24(3):150-157. https://doi.org/10.1093/labmed/24.3.150
47.
Currier MG, Lee S, Stobart CC, Hotard AL, Villenave R, Meng J, Pretto CD, Shields MD, Nguyen MT, Todd SO, et al. EGFR Interacts with the Fusion Protein of Respiratory Syncytial Virus Strain 2-20 and Mediates Infection and Mucin Expression. PLoS Pathog. 12(5):e1005622. https://doi.org/10.1371/journal.ppat.1005622
48.
Tan B, Wu L, Yang X, Li B, Zhang W, Lei Y, Li Y, Yang G, Chen J, Chen G, et al. 2016. Isolation and characterization of adenoviruses infecting endangered golden snub-nosed monkeys (Rhinopithecus roxellana). Virol J. 13(1): https://doi.org/10.1186/s12985-016-0648-6
49.
Yang T, Li S, Zhang X, Pang X, Lin Q, Cao J. 2015. Resveratrol, sirtuins, and viruses. Rev. Med. Virol.. 25(6):431-445. https://doi.org/10.1002/rmv.1858
50.
Gagliardi TB, Criado MF, Proença-Módena JL, Saranzo AM, Iwamoto MA, de Paula FE, Cardoso RS, Delcaro LS, Silva ML, Câmara AA, et al. 2017. Syncytia Induction by Clinical Isolates of Human Respiratory Syncytial Virus A. Intervirology. 60(1-2):56-60. https://doi.org/10.1159/000480014
51.
Amatore D, Sgarbanti R, Aquilano K, Baldelli S, Limongi D, Civitelli L, Nencioni L, Garaci E, Ciriolo MR, Palamara AT. 2015. Influenza virus replication in lung epithelial cells depends on redox?sensitive pathways activated by NOX4 ?derived ROS. Cell Microbiol. 17(1):131-145. https://doi.org/10.1111/cmi.12343
52.
Castells E, George VG, Hierholzer JC. 1990. NCI-H292 as an alternative cell line for the isolation and propagation of the human paramyxoviruses. Archives of Virology. 115(3-4):277-288. https://doi.org/10.1007/bf01310536
53.
Bianco A, Sethi S, Allen J, Knight R, Spiteri M. 1998. Th2 cytokines exert a dominant influence on epithelial cell expression of the major group human rhinovirus receptor, ICAM-1. 12(3):619-626. https://doi.org/10.1183/09031936.98.12030619
54.
Mall AS, Habte H, Mthembu Y, Peacocke J, de Beer C. 2017. Mucus and Mucins: do they have a role in the inhibition of the human immunodeficiency virus?. Virol J. 14(1): https://doi.org/10.1186/s12985-017-0855-9
55.
Elion GB, Furman PA, Fyfe JA, Miranda Pd, Beauchamp L, Schaeffer HJ. 1977. Selectivity of action of an antiherpetic agent, 9-(2-hydroxyethoxymethyl)guanine. Proceedings of the National Academy of Sciences. 74(12):5716-5720. https://doi.org/10.1073/pnas.74.12.5716
56.
Rima BK, Davidson WB, Martin SJ. 1977. The Role of Defective Interfering Particles in Persistent Infection of Vero Cells by Measles Virus. Journal of General Virology. 35(1):89-97. https://doi.org/10.1099/0022-1317-35-1-89
57.
Gouma S, ten Hulscher HI, Schurink-van ?t Klooster TM, de Melker HE, Boland GJ, Kaaijk P, van Els CA, Koopmans MP, van Binnendijk RS. 2016. Mumps-specific cross-neutralization by MMR vaccine-induced antibodies predicts protection against mumps virus infection. Vaccine. 34(35):4166-4171. https://doi.org/10.1016/j.vaccine.2016.06.063
58.
Sanders BP, Oakes IdlR, van Hoek V, Liu Y, Marissen W, Minor PD, Wimmer E, Schuitemaker H, Custers JH, Macadam A, et al. 2015. Production of high titer attenuated poliovirus strains on the serum-free PER.C6® cell culture platform for the generation of safe and affordable next generation IPV. Vaccine. 33(48):6611-6616. https://doi.org/10.1016/j.vaccine.2015.10.091
59.
Kang JY, Lee DK, Ha NJ, Shin HS. 2015. Antiviral effects of Lactobacillus ruminis SPM0211 and Bifidobacterium longum SPM1205 and SPM1206 on rotavirus-infected Caco-2 cells and a neonatal mouse model. J Microbiol.. 53(11):796-803. https://doi.org/10.1007/s12275-015-5302-2
60.
Mangala Prasad V, Klose T, Rossmann MG. Assembly, maturation and three-dimensional helical structure of the teratogenic rubella virus. PLoS Pathog. 13(6):e1006377. https://doi.org/10.1371/journal.ppat.1006377
61.
Venturi G, Zammarchi L, Fortuna C, Remoli ME, Benedetti E, Fiorentini C, Trotta M, Rizzo C, Mantella A, Rezza G, et al. 2016. An autochthonous case of Zika due to possible sexual transmission, Florence, Italy, 2014. 21(8): https://doi.org/10.2807/1560-7917.es.2016.21.8.30148
62.
Criscuolo E, Clementi N, Mancini N, Burioni R, Miduri M, Castelli M, Clementi M. 2018. Synergy evaluation of anti-Herpes Simplex Virus type 1 and 2 compounds acting on different steps of virus life cycle. Antiviral Research. 15171-77. https://doi.org/10.1016/j.antiviral.2018.01.009
63.
Richard AS, Shim B, Kwon Y, Zhang R, Otsuka Y, Schmitt K, Berri F, Diamond MS, Choe H. 2017. AXL-dependent infection of human fetal endothelial cells distinguishes Zika virus from other pathogenic flaviviruses. Proc Natl Acad Sci USA. 114(8):2024-2029. https://doi.org/10.1073/pnas.1620558114
64.
Kim YY, Jee HJ, Um J, Kim YM, Bae SS, Yun J. 2017. Cooperation between p21 and Akt is required for p53-dependent cellular senescence. Aging Cell. 16(5):1094-1103. https://doi.org/10.1111/acel.12639
65.
Plotkin S. 2015. The history of vaccination against cytomegalovirus. Med Microbiol Immunol. 204(3):247-254. https://doi.org/10.1007/s00430-015-0388-z
66.
Mintz L, Drew WL. 1980. Relation of Culture Site to the Recovery of Nonpolio Enteroviruses. 74(3):324-326. https://doi.org/10.1093/ajcp/74.3.324
67.
Baker DA, Kleger B, Plotkin SA. 1979. A Comparison of Herpes Simplex Virus (HSV) Typing Using Direct Fluorescent Antibody (DFA) and Chick Embryo Plaque (CEP) Assays. Lab Med. 10(9):559-561. https://doi.org/10.1093/labmed/10.9.559
68.
Perelygina L, Plotkin S, Russo P, Hautala T, Bonilla F, Ochs HD, Joshi A, Routes J, Patel K, Wehr C, et al. 2016. Rubella persistence in epidermal keratinocytes and granuloma M2 macrophages in patients with primary immunodeficiencies. Journal of Allergy and Clinical Immunology. 138(5):1436-1439.e11. https://doi.org/10.1016/j.jaci.2016.06.030
69.
Sousa FH, Casanova V, Findlay F, Stevens C, Svoboda P, Pohl J, Proudfoot L, Barlow PG. 2017. Cathelicidins display conserved direct antiviral activity towards rhinovirus. Peptides. 9576-83. https://doi.org/10.1016/j.peptides.2017.07.013
70.
Shariff S, Shelfoon C, Holden NS, Traves SL, Wiehler S, Kooi C, Proud D, Leigh R. 2017. Human Rhinovirus Infection of Epithelial Cells Modulates Airway Smooth Muscle Migration. Am J Respir Cell Mol Biol. 56(6):796-803. https://doi.org/10.1165/rcmb.2016-0252oc
71.
Corry J, Johnson SM, Cornwell J, Peeples ME. 2016. Preventing Cleavage of the Respiratory Syncytial Virus Attachment Protein in Vero Cells Rescues the Infectivity of Progeny Virus for Primary Human Airway Cultures. J. Virol.. 90(3):1311-1320. https://doi.org/10.1128/jvi.02351-15
72.
Zahoor MA, Khurshid M, Qureshi R, Naz A, Shahid M. 2016. Cell culture-based viral vaccines: current status and future prospects. Future Virology. 11(7):549-562. https://doi.org/10.2217/fvl-2016-0006