Merck
Different outcome of sarcoglycan missense mutation between human and mouse.
PloS one (2018-01-24)
Sara F Henriques, Cécile Patissier, Nathalie Bourg, Chiara Fecchio, Doriana Sandona, Justine Marsolier, Isabelle Richard
ZUSAMMENFASSUNG
Sarcoglycanopathies are rare autosomic limb girdle muscular dystrophies caused by mutations in one of the genes coding for sarcoglycan (α, β, δ, and γ-sarcoglycans). Sarcoglycans form a complex, which is an important part of the dystrophin-associated glycoprotein complex that protects sarcolemma against muscle contraction-induced damages. Absence of one of the sarcoglycan at the plasma membrane induces the disappearance of the whole complex and perturbs muscle fiber membrane integrity. We previously demonstrated that point mutations in the human sarcoglycan genes affects the folding of the corresponding protein, which is then retained in the endoplasmic reticulum by the protein quality control and prematurely degraded by the proteasome. Interestingly, modulation of the quality control using pharmacological compounds allowed the rescue of the membrane localization of the mutated sarcoglycan. Two previously generated mouse models, knock-in for the most common sarcoglycan mutant, R77C α-sarcoglycan, failed in reproducing the dystrophic phenotype observed in human patients. Based on these results and the need to test therapies for these fatal diseases, we decided to generate a new knock-in mouse model carrying the missense mutation T151R in the β-sarcoglycan gene since this is the second sarcoglycan protein with the most frequently reported missense mutations. Muscle analysis, performed at the age of 4 and 9-months, showed the presence of the mutated β-sarcoglycan protein and of the other components of the dystrophin-associated glycoprotein complex at the muscle membrane. In addition, these mice did not develop a dystrophic phenotype, even at a late stage or in condition of stress-inducing exercise. We can speculate that the absence of phenotype in mouse may be due to a higher tolerance of the endoplasmic reticulum quality control for amino-acid changes in mice compared to human.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-SGCB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.