Merck
A novel TNF receptor family member binds TWEAK and is implicated in angiogenesis.
Immunity (2001-12-01)
S R Wiley, L Cassiano, T Lofton, T Davis-Smith, J A Winkles, V Lindner, H Liu, T O Daniel, C A Smith, W C Fanslow
ZUSAMMENFASSUNG
TWEAK is a member of the TNF ligand family that induces angiogenesis in vivo. We report cloning of a receptor for TWEAK (TweakR) from a human umbilical vein endothelial cell (HUVEC) library. The mature form of TweakR has only one hundred and two amino acids and six cysteine residues in its extracellular region. Five different assays demonstrate TWEAK-TweakR binding, and the interaction affinity constant (Kd) is within a physiologically relevant range of 2.3 +/- 0.1 nM. The TweakR cytoplasmic domain binds TRAFs 1, 2, and 3. Cross-linking of TweakR induces HUVEC growth, and mRNA levels are upregulated in vitro by a variety of agents and in vivo following arterial injury. Soluble TweakR inhibits endothelial cell migration in vitro and corneal angiogenesis in vivo.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
TWEAK Receptor human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.