Merck
Adipocytes promote ovarian cancer chemoresistance.
Scientific reports (2019-09-19)
Jiang Yang, Munir M Zaman, Iliyan Vlasakov, Roopali Roy, Lan Huang, Camilia R Martin, Steven D Freedman, Charles N Serhan, Marsha A Moses
ZUSAMMENFASSUNG
Ovarian cancer (OvCa), while accounting for only 3% of all women's cancer, is the fifth leading cause of cancer death among women. One of the most significant obstacles to successful OvCa treatment is chemoresistance. The current lack of understanding of the driving mechanisms underlying chemoresistance hinders the development of effective therapeutics against this obstacle. Adipocytes are key components of the OvCa microenvironment and have been shown to be involved in OvCa cell proliferation, however, little is known about their impact on OvCa chemoresistance. In the current study, we found that adipocytes, of both subcutaneous and visceral origin, secrete factors that enhance the resistance of OvCa cells against chemotherapeutic drugs by activating the Akt pathway. Importantly, we have demonstrated that secreted lipids mediate adipocyte-induced chemoresistance. Through a comprehensive lipidomic analysis, we have identified this chemo-protective lipid mediator as arachidonic acid (AA). AA acts on OvCa cells directly, not through its downstream derivatives such as prostaglandins, to activate Akt and inhibit cisplatin-induced apoptosis. Taken together, our study has identified adipocytes and their secreted AA as important mediators of OvCa chemoresistance. Strategies that block the production of AA from adipocytes or block its anti-apoptotic function may potentially inhibit chemoresistance in OvCa patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
PD 146176, ≥98% (HPLC), solid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.