Merck
Over-expression of EGFR regulated by RARA contributes to 5-FU resistance in colon cancer.
Aging (2020-01-04)
Xin-Yue Gu, Yang Jiang, Ming-Qi Li, Peng Han, Yan-Long Liu, Bin-Bin Cui
ZUSAMMENFASSUNG
A promising new strategy for cancer therapy is to target the autophagic pathway. However, comprehensive characterization of autophagy genes and their clinical relevance in cancer is still lacking. Here, we systematically characterized alterations of autophagy genes in multiple cancer lines by analyzing data from The Cancer Genome Atlas and CellMiner database. Interactions between autophagy genes and clinically actionable genes (CAGs) were identified by analyzing co-expression, protein-protein interactions (PPIs) and transcription factor (TF) data. A key subnetwork was identified that included 18 autophagy genes and 22 CAGs linked by 28 PPI pairs and 1 TF-target pair, which was EGFR targeted by RARA. Alterations in the expression of autophagy genes were associated with patient survival in multiple cancer types. RARA and EGFR were associated with worse survival in colorectal cancer patients. The regulatory role of EGFR in 5-FU resistance was validated in colon cancer cells in vivo and in vitro. EGFR contributed to 5-FU resistance in colon cancer cells through autophagy induction, and EGFR overexpression in 5-FU resistant colon cancer was regulated by RARA. The present study provides a comprehensive analysis of autophagy in different cancer cell lines and highlights the potential clinical utility of targeting autophagy genes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human EGFR
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
MISSION® esiRNA, targeting human RARA
Preise und Verfügbarkeit sind derzeit nicht verfügbar.