Merck
  • Benzylimidazolines as h5-HT1B/1D serotonin receptor ligands: a structure-affinity investigation.
Benzylimidazolines as h5-HT1B/1D serotonin receptor ligands: a structure-affinity investigation.
Journal of medicinal chemistry (1998-06-19)
H Law, M Dukat, M Teitler, D K Lee, L Mazzocco, R Kamboj, V Rampersad, T Prisinzano, R A Glennon
ZUSAMMENFASSUNG
Benzylimidazolines may represent a class of 5-HT1D ligands that has yet to be exploited. On the basis of a previous report that the 2-(substituted-benzyl)imidazoline alpha-adrenergic agonist oxymetazoline (8) binds with high affinity at calf brain 5-HT1D receptors, we explored the structure-affinity relationships of a series of related derivatives. Each of the aromatic substituents was removed and then reinstated in a systematic manner to determine the influence of the individual substituents on binding. It was found that all of the aromatic substituents of 8 act in concert to impart high affinity. However, although the 3-hydroxy group could be removed without significantly reducing affinity for h5-HT1D (i.e., human 5-HT1Dalpha) receptors, this modification reduced h5-HT1B (i.e., human 5-HT1Dbeta) receptor affinity by nearly 50-fold. The 2, 6-dimethyl groups also contribute to binding but seem to play a greater role for h5-HT1B binding than h5-HT1D binding. With the appropriate structural modifications, several compounds were identified that display 20- to >100-fold selectivity for h5-HT1D versus h5-HT1B receptors. Preliminary functional data suggest that these compounds behave as agonists. Given that 5-HT1D agonists are currently being explored for their antimigraine action and that activation of h5-HT1B receptors might be associated with cardiovascular side effects, h5-HT1D-selective agents may offer a new lead for the development of therapeutically efficacious agents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oxymetazolin -hydrochlorid, ≥99%, solid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.