Merck
  • Hippocampal Mrp8/14 signaling plays a critical role in the manifestation of depressive-like behaviors in mice.
Hippocampal Mrp8/14 signaling plays a critical role in the manifestation of depressive-like behaviors in mice.
Journal of neuroinflammation (2018-09-06)
Hong Gong, Wen-Jun Su, Zhi-Yong Cao, Yong-Jie Lian, Wei Peng, Yun-Zi Liu, Yi Zhang, Lin-Lin Liu, Ran Wu, Bo Wang, Ting Zhang, Yun-Xia Wang, Chun-Lei Jiang
ZUSAMMENFASSUNG
Depression is one of the most common mental disorders characterized mainly by low mood and loss of interest or pleasure. About a third of patients with depression do not respond to classic antidepressant treatments. Recent evidence suggests that Mrp8/14 (myeloid-related protein 8/14) plays a crucial role in cognitive dysfunction and neuroinflammatory diseases, yet its role in mood regulation remains largely uninvestigated. In the present work, we explored the potential role of Mrp8/14 in the progression of depression. After 4 weeks of chronic unpredictable mild stress (CUMS), depressive-like symptoms and Mrp8/14 were determined. To verify the effects of Mrp8/14 on depressive-like behaviors, the inhibitor TAK-242 and recombinant Mrp8/14 were used. Furthermore, the molecular mechanisms in Mrp8/14-induced behavioral and biological changes were examined in vivo and ex vivo. Four-week CUMS contributed to the development of depressive symptoms. Mrp8 and Mrp14 were upregulated in the hippocampus and serum after exposure to CUMS. Pharmacological inhibition of Mrp14 attenuated CUMS-induced TLR4/NF-κB signaling activation and depressive-like behaviors. Furthermore, central administration of recombinant Mrp8, Mrp14, and Mrp8/14 resulted in neuroinflammation and depressive-like behaviors. Mrp8/14-provoked proinflammatory effects and depressive-like behaviors were improved by pretreatment with a TLR4 inhibitor. Moreover, pharmacological inhibition of TLR4 reduced the release of nitric oxide and reactive oxygen species in Mrp8/14-activated BV2 microglia. These data suggest that the hippocampal Mrp8/14-TLR4-mediated neuroinflammation contributes to the development of depressive-like behaviors. Targeting the Mrp8/14 may be a novel promising antidepressant approach.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Paquinimod, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.