Merck
  • 1H and 13C NMR assignments for two oxaphenalenones bacillosporin C and D from the mangrove endophytic fungus SBE-14.
1H and 13C NMR assignments for two oxaphenalenones bacillosporin C and D from the mangrove endophytic fungus SBE-14.
Magnetic resonance in chemistry : MRC (2007-03-21)
Zhiyong Guo, Changlun Shao, Zhigang She, Xiaoling Cai, Fan Liu, L L P Vrijimoed, Yongcheng Lin
ZUSAMMENFASSUNG
The complete (1)H and (13)C NMR assignments are reported for the novel natural product Bacillosporin D together with the known compound Bacillosporin C. These compounds containing seven rings were isolated from the mangrove endophytic fungus SBE-14 from the South China Sea. 1D and 2D NMR experiments, including COSY, HMQC and HMBC were used to the determination of the structures and NMR assignments. It is proposed that 1 was biogenetically produced by transforming 2. Transforming a lactone to an anhydride is unusual in nature.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Perinaphthenon, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.