Merck
  • Balearic adults have low intakes of fruits and vegetables compared with the dietary guidelines for adults in Spain.
Balearic adults have low intakes of fruits and vegetables compared with the dietary guidelines for adults in Spain.
Nutrition research (New York, N.Y.) (2013-03-20)
Rogelio Salas, Maria del Mar Bibiloni, Maria Elisa Zapata, Josep Lluis Coll, Antoni Pons, Josep A Tur
ZUSAMMENFASSUNG
Nutrition is recognized as one of the major health determinants, and so a healthy diet may contribute to the delay or prevention of an important number of chronic diseases. The aim of this study was to assess the current food consumption habits of the Balearic Islands adult population. A cross-sectional nutritional survey was carried out in the Balearic Islands, Spain (2009-2010). A random sample (n = 1388) of the adult population (16-65 years) was interviewed. Dietary habits were assessed by means of a semiquantitative food frequency questionnaire, and consumption results were compared with dietary guidelines for the Spanish population. According to the dietary guidelines for the Spanish population, only consumption levels of milk and dairy products, nuts, potatoes, cereals, bread, and water were adequate. Intakes of fruit, vegetables, olive oil, eggs, and pulses were below the recommendation levels. These findings indicate that, to fulfill the dietary guidelines for the Spanish population, the Balearic Islands adult population should decrease their consumption of meat, sugar and cakes, buns, butter/margarine, and soft drinks and increase their consumption of olive oil, fruits, vegetables, fish, and pulses.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Olivenöl, highly refined, low acidity
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Olivenöl, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Olivenöl, tested according to Ph. Eur.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.