- [Oxidation of alpha-methylstyrene by Pseudomonas cultures].
[Oxidation of alpha-methylstyrene by Pseudomonas cultures].
Mikrobiologiia (1985-01-01)
D B Dzhusupova, B P Baskunov, L A Golovleva, R M Alieva, A N Ilialetdinov
PMID3925301
ZUSAMMENFASSUNG
The work was aimed at studying alpha-methylstyrene oxidation by three active Pseudomonas cultures. P. acidovorans 9 oxidized alpha-methylstyrene only to acetophenone whereas two P. aeruginosa cultures assimilated the compound entirely. Analysis of the intermediate products and enzymes catalyzing the aromatic ring cleavage suggests that these strains oxidize alpha-methylstyrene via a new unknown pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Preise und Verfügbarkeit sind derzeit nicht verfügbar.