Merck
  • Stereoselective inactivation of Torpedo californica acetylcholinesterase by isomalathion: inhibitory reactions with (1R)- and (1S)-isomers proceed by different mechanisms.
Stereoselective inactivation of Torpedo californica acetylcholinesterase by isomalathion: inhibitory reactions with (1R)- and (1S)-isomers proceed by different mechanisms.
Chemical research in toxicology (2003-08-20)
Jonathan A Doorn, Charles M Thompson, Robert B Christner, Rudy J Richardson
ZUSAMMENFASSUNG
The present study was undertaken to test the hypothesis that acetylcholinesterase (AChE) inhibition by isomalathion stereoisomers proceeds with different primary leaving groups for (1R)- and (1S)-isomers. Consistent with results obtained with enzyme from other species, AChE from Torpedo californica (TcAChE) was stereoselectively inhibited by isomalathion isomers with the (1R,3R)-isomer exhibiting greater potency than (1S,3S)-isomalathion. TcAChE modified by (1R)-isomers readily reactivated in the presence of 2-pralidoxime methiodide (2-PAM), whereas enzyme inhibited by (1S)-isomalathions was intractable toward reactivation. Computer-based molecular modeling showed that the ligand positioned as the primary leaving group was diethyl thiosuccinyl for (1R)-isomers and thiomethyl for (1S)-isomalathions. Mass spectral analysis revealed that inhibition of TcAChE by (1R)-isomers resulted in an O,S-dimethyl phosphate adduct, as expected from expulsion of the diethyl thiosuccinyl ligand. In contrast, inactivation of the enzyme by (1S)-isomalathions yielded an O-methyl phosphate adduct, consistent with initial loss of thiomethyl followed by displacement of the diethyl thiosuccinyl group. The findings demonstrate that the inhibitory reactions of TcAChE with (1R)- and (1S)-isomalathions proceed by different mechanisms involving distinct primary leaving groups.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Malathion-Unreinheit A, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.