Merck
5-lipoxygenase is a direct target of miR-19a-3p and miR-125b-5p.
Journal of immunology (Baltimore, Md. : 1950) (2015-01-16)
Saskia Busch, Eileen Auth, Friederike Scholl, Sabine Huenecke, Ulrike Koehl, Beatrix Suess, Dieter Steinhilber
ZUSAMMENFASSUNG
5-Lipoxygenase (5-LO) is the key enzyme in leukotriene biosynthesis. Leukotrienes are mediators of the innate immune system and inflammatory processes, and they might also be involved in cancer development. MicroRNAs (miRNAs) are important translational regulators and have been shown to be involved in development, differentiation, and cancer. Unraveling the miRNA network is important for understanding the cellular regulation processes. We identified two new miRNAs, miR-19a-3p and miR-125b-5p, regulating 5-LO and confirmed direct interaction by reporter gene assays. Furthermore, we investigated the regulation of 5-LO by these two miRNAs in several cell types. Inhibition of both miRNAs by antagomirs during differentiation of the myeloid cell line Mono Mac 6 led to a significant increase in 5-LO protein expression. Stimulation of human T lymphocytes with PHA resulted in a strong downregulation of 5-LO mRNA expression and in the induction of miR-19a-3p. The inhibition of miR-19a-3p with an antagomir led to a significant increase in 5-LO mRNA expression in T lymphocytes. Taken together, our data reveal that miR-19a-3p and miR-125b-5p target 5-LO in a cell type- and stimulus-specific manner.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Essigsäure, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Essigsäure, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Guanidinthiocyanat, Molecular Biology, ≥99%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
USP
Eisessig, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)