Merck
  • Low-molecular-mass secretome profiling identifies HMGA2 and MIF as prognostic biomarkers for oral cavity squamous cell carcinoma.
Low-molecular-mass secretome profiling identifies HMGA2 and MIF as prognostic biomarkers for oral cavity squamous cell carcinoma.
Scientific reports (2015-07-04)
Kai-Ping Chang, Shih-Jie Lin, Shiau-Chin Liu, Jui-Shan Yi, Kun-Yi Chien, Lang-Ming Chi, Huang-Kai Kao, Ying Liang, Yu-Tsun Lin, Yu-Sun Chang, Jau-Song Yu
ZUSAMMENFASSUNG
The profiling of cancer cell secretomes is considered to be a good strategy for identifying cancer-related biomarkers, but few studies have focused on identifying low-molecular-mass (LMr) proteins (<15 kDa) in cancer cell secretomes. Here, we used tricine-SDS-gel-assisted fractionation and LC-MS/MS to systemically identify LMr proteins in the secretomes of five oral cavity squamous cell carcinoma (OSCC) cell lines. Cross-matching of these results with nine OSCC tissue transcriptome datasets allowed us to identify 33 LMr genes/proteins that were highly upregulated in OSCC tissues and secreted/released from OSCC cells. Immunohistochemistry and quantitative real-time PCR were used to verify the overexpression of two candidates, HMGA2 and MIF, in OSCC tissues. The overexpressions of both proteins were associated with cervical metastasis, perineural invasion, deeper tumor invasion, higher overall stage, and a poorer prognosis for post-treatment survival. Functional assays further revealed that both proteins promoted the migration and invasion of OSCC cell lines in vitro. Collectively, our data indicate that the tricine-SDS-gel/LC-MS/MS approach can be used to efficiently identify LMr proteins from OSCC cell secretomes, and suggest that HMGA2 and MIF could be potential tissue biomarkers for OSCC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
3,3′-Diaminobenzidin -tetrahydrochlorid, tablet, 10 mg substrate per tablet
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Anti-Aktin-Antikörper, Klon C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG