Merck
  • Physiology and proteomics research on the leaves of ancient Platycladus orientalis (L.) during winter.
Physiology and proteomics research on the leaves of ancient Platycladus orientalis (L.) during winter.
Journal of proteomics (2015-07-15)
Sheng Zhang, Lingling Zhang, Yongyu Chai, Fei Wang, Yiming Li, Li Su, Zhong Zhao
ZUSAMMENFASSUNG
Ancient trees have an important value in humanities and history, and also have an important scientific value in the investigation of the decline and senescence mechanisms. Thus, we conducted an environmental stress study using ancient trees. To evaluate age-dependent changes in physiology and the leaf proteome, we assessed the low-temperature stress responses of 20 ± 5-, 500 ± 100- and 1200 ± 200-year-old Platycladus orientalis (L.) samples obtained outdoors during winter. Several physiological parameters were evaluated. Leaf proteomes were obtained using two-dimensional electrophoresis gels, and 77 protein spots were identified successfully using MALDI TOF/TOF MS/MS. The majority of the identified protein species were classified into functional categories including defense/stress-related, energy and carbohydrate metabolism, photosynthesis, and hormone-related functions. A general reduction in the abundance of protein species was observed as the age of the studied trees increased; reduction in photosynthesis and defense/stress-related categories were particularly apparent in the leaves of ancient trees. However, the number of protein species with functions in energy and carbohydrate metabolism increased with age. An increase in the abundance of lipid metabolism and hormone-related protein species was a primary characteristic of the leaves of ancient trees under low-temperature stress during winter. These results improve our understanding of the biochemical mechanisms of stress responses in ancient trees. Low temperature is the most common meteorological challenge in the study area. For evergreen plants, low-temperature stress has a great impact on the leaves of ancient P. orientalis. Thus, we conducted an environmental stress study using ancient trees. Recently, various studies were carried out in ancient trees. However, no information is available on the molecular mechanisms of defense to low-temperature stress in ancient trees. Therefore, this original study comprises the following differential proteomic analysis of ancient P. orientalis: (1) age-dependent changes in the physiology and leaf proteome are evaluated under low-temperature stress to (2) understand the differences in metabolic responses between ancient and adult trees under low-temperature stress during winter. This analysis will provide an understanding of the complex physiological changes that occur in ancient trees. The results suggest that certain identified proteins can be used as markers of low-temperature stress in ancient P. orientalis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Saccharose, ACS reagent
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
SAFC
Glycin
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
DL-Methionin, ≥99%