Merck
  • International Regulations of Propolis Quality: Required Assays do not Necessarily Reflect their Polyphenolic-Related In Vitro Activities.
International Regulations of Propolis Quality: Required Assays do not Necessarily Reflect their Polyphenolic-Related In Vitro Activities.
Journal of food science (2015-05-07)
Raquel Bridi, Gloria Montenegro, Gabriel Nuñez-Quijada, Ady Giordano, Maria Fernanda Morán-Romero, Isaac Jara-Pezoa, Hernán Speisky, Elias Atala, Camilo López-Alarcón
ZUSAMMENFASSUNG
Propolis has been proposed as a polyphenolic-rich natural product potentially able to be used for human consumption or even for medicinal proposes. To guarantee a minimum phenolic and flavonoid content and as consequence of their related-biological activities, international requirements of propolis quality are commonly applied. In this work we assessed phenolic and flavonoid contents of propolis; the antioxidant capacity (toward peroxyl radicals and hypochlorous acid); the ability to generate nitric oxide (NO); and, finally the antimicrobial activity of 6 propolis samples from the VI region of Chile. Our results show that the total phenolic and flavonoid content of propolis samples are not always in agreement with their polyphenolic-associated in vitro activities. For example, P03 and P06 samples showed the lowest (25 ± 4 GAE/g propolis) and the highest (105 ± 3 GAE/g propolis) total phenolic content, respectively. This was in agreement with flavonoid content and their Oxygen Radical Absorbance Capacity (ORAC) activity. However, this dependence was not observed toward HOCl, NO release and antimicrobial activity. Based on our results, we consider that, in order to guarantee the antioxidant or antimicrobial in vitro effects, the international regulations of propolis quality should contemplate the convenience of incorporating other simple analytical test such as ORAC or antimicrobial tests.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2,2-Diphenyl-1-Picrylhydrazyl
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchroman-2-carbonsäure, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumphosphat Dihydrat, BioUltra, Molecular Biology, ≥99.0% (T)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumphosphat, ReagentPlus®, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumphosphat, BioReagent, Molecular Biology, anhydrous, ≥98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2,2′-Azo-bis(2-methylpropionamidin) -dihydrochlorid, powder or granules, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumphosphat, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Fluorescein, free acid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumnitrit, 99.999% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aluminiumchlorid, anhydrous, powder, 99.999% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumphosphat, meets USP testing specifications, anhydrous
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumphosphat monobasisch -Lösung, BioUltra, 5 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aluminiumchlorid, 99.99% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumphosphat, BioXtra, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumphosphat, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aluminiumchlorid, powder, 99.99% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumphosphat monobasisch-16O4, 99.9 atom % 16O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.