Merck
  • Detection and Quantification of Gluten during the Brewing and Fermentation of Beer Using Antibody-Based Technologies.
Detection and Quantification of Gluten during the Brewing and Fermentation of Beer Using Antibody-Based Technologies.
Journal of food protection (2015-06-04)
Rakhi Panda, Hans F Zoerb, Chung Y Cho, Lauren S Jackson, Eric A E Garber
ZUSAMMENFASSUNG
In 2013 the U.S. Food and Drug Administration (FDA) defined the term ''gluten-free'' and identified a gap in the analytical methodology for detection and quantification of gluten in foods subjected to fermentation and hydrolysis. To ascertain the ability of current enzyme-linked immunosorbent assays (ELISAs) to detect and quantify gluten in fermented and hydrolyzed products, sorghum beer was spiked in the initial phases of production with 0, 20, and 200 μg/ml wheat gluten, and samples were collected throughout the beer production process. The samples were analyzed using five sandwich ELISAs and two competitive ELISAs and by sodium dodecyl sulfate-polyacrylamide gel electrophoresis with Western analysis employing four antibodies (MIoBS, R5, G12, and Skerritt). The sensitivity of the MIoBS ELISA (0.25 ppm) enabled the reliable detection of gluten throughout the manufacturing process, including fermentation, when the initial concentration of 20 μg/ml dropped to 2 μg/ml. The R5 antibody-based and G12 antibody-based sandwich ELISAs were unable to reliably detect gluten, initially at 20 μg/ml, after the onset of production. The Skerritt antibody-based sandwich ELISA overestimated the gluten concentration in all samples. The R5 antibody-based and G12 antibody-based competitive ELISAs were less sensitive than the sandwich ELISAs and did not provide accurate results for quantifying gluten concentration. The Western analyses were able to detect gluten at less than 5 μg/ml in the samples and confirmed the results of the ELISAs. Although further research is necessary before all problems associated with detection and quantification of hydrolyzed and fermented gluten are resolved, the analytical methods recommended by the FDA for regulatory samples can detect ≥ 20 μg/ml gluten that has undergone brewing and fermentation processes associated with the manufacture of beer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
TWEEN® 20, BioXtra, viscous liquid
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
L-Lysin -monohydrochlorid, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Phosphorsäure -Lösung, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Phosphorsäure, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphatgepufferte Kochsalzlösung -Lösung, powder, pH 7.4, for preparing 5 L solutions
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Gluten from wheat
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
L-Lysin -monohydrochlorid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Phosphorsäure, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Phosphorsäure, BioReagent, suitable for insect cell culture, 85%