Merck
  • Neural correlates of migration: activation of hypothalamic clock(s) in and out of migratory state in the blackheaded bunting (Emberiza melanocephala).
Neural correlates of migration: activation of hypothalamic clock(s) in and out of migratory state in the blackheaded bunting (Emberiza melanocephala).
PloS one (2013-11-10)
Ashutosh Rastogi, Yatinesh Kumari, Sangeeta Rani, Vinod Kumar
ZUSAMMENFASSUNG
Many vertebrates distinguish between short and long day lengths using suprachiasmatic nuclei (SCN). In birds particular, the mediobasal hypothalamus (MBH) is suggested to be involved in the timing of seasonal reproduction. This study investigated the response of SCN and MBH to a single long day, and the role of MBH in induction of the migratory phenotype in night-migratory blackheaded buntings. Experiment 1 immunocytochemically measured c-fos in the SCN, and c-fos, vasoactive intestinal peptide (VIP) and neuropeptide Y (NPY) in the MBH of buntings exposed to a 20 h light period. Long light period induced significantly stronger c-fos expression, measured as number of c-fos-like immunoreactive (c-fos-lir) cells, in MBH, but not in the SCN. Within the MBH, c-fos-lir cells were significantly denser in the inferior hypothalamic nucleus (IH) and infundibular nucleus (IN), but not in the dorsomedial hypothalamus (DMH). IH and IN also had significantly increased number of VIP and NPY labeled cells. DMH had significantly increased number of VIP labeled cells only. Experiment 2 assayed c-fos, VIP and NPY immunoreactivities in the middle of day and night in the MBH of buntings, after seven long days (day active, non-migratory state) and after seven days of Zugunruhe (night active, migratory state) in long days. In the migratory state, the number of c-fos-lir cells was significantly greater only in DMH; VIP-lir cells were denser in all three MBH regions suggesting enhanced light sensitivity at night. The denser NPY-lir cells only in IN in the non-migratory state were probably due to premigratory hyperphagia. In buntings, SCN may not be involved in the photoperiod-induced seasonal responses. MBH contains the seasonal clock sensitive to day length. VIP and NPY are parts of the neuroendocrine mechanism(s) involved, respectively, in sensing and translating the photoperiodic message in a seasonal response.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Neuropeptide Y (NPY) antibody produced in rabbit, whole antiserum
Preise und Verfügbarkeit sind derzeit nicht verfügbar.