Merck
  • Identification of residues in the N-terminal PAS domains important for dimerization of Arnt and AhR.
Identification of residues in the N-terminal PAS domains important for dimerization of Arnt and AhR.
Nucleic acids research (2011-01-20)
Nan Hao, Murray L Whitelaw, Keith E Shearwin, Ian B Dodd, Anne Chapman-Smith
ZUSAMMENFASSUNG
The basic helix-loop-helix (bHLH).PAS dimeric transcription factors have crucial roles in development, stress response, oxygen homeostasis and neurogenesis. Their target gene specificity depends in part on partner protein choices, where dimerization with common partner Aryl hydrocarbon receptor nuclear translocator (Arnt) is an essential step towards forming active, DNA binding complexes. Using a new bacterial two-hybrid system that selects for loss of protein interactions, we have identified 22 amino acids in the N-terminal PAS domain of Arnt that are involved in heterodimerization with aryl hydrocarbon receptor (AhR). Of these, Arnt E163 and Arnt S190 were selective for the AhR/Arnt interaction, since mutations at these positions had little effect on Arnt dimerization with other bHLH.PAS partners, while substitution of Arnt D217 affected the interaction with both AhR and hypoxia inducible factor-1α but not with single minded 1 and 2 or neuronal PAS4. Arnt uses the same face of the N-terminal PAS domain for homo- and heterodimerization and mutational analysis of AhR demonstrated that the equivalent region is used by AhR when dimerizing with Arnt. These interfaces differ from the PAS β-scaffold surfaces used for dimerization between the C-terminal PAS domains of hypoxia inducible factor-2α and Arnt, commonly used for PAS domain interactions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
YH439, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.