Merck
Synthetic and naturally occurring hydrazines as possible cancer causative agents.
Cancer research (1975-12-01)
B Toth
ZUSAMMENFASSUNG
The various synthetic substituted hydrazines, which cause tumors in animals, are briefly enumerated. To date, 19 of them have proved to be tumorigenic in animals. A number of these chemicals are found today in the environment, in industry, in agriculture, and in medicine, and the human population is exposed to a certain degree to some of them. Hydrazine also occurs in nature in tobacco and tobacco smoke. The three other naturally occurring hydrazine compounds are N-methyl-N-formylhydrazine, which occurs in the wild edible mushroom, Gyromitra esculenta, and beta-N-[gamma-L(+)-glutamyl]-4-hydroxymethylphenylhydrazine and 4-hydroxymethylphenylhydrazine, which are found in the commonly eaten cultivated mushroom, Agaricus bisporus. Tumorigenesis studies with the naturally occurring hydrazines are in progress.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,1-Dimethylhydrazin, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.