Merck
The Nedd4-2/Ndfip1 axis is a negative regulator of IgE-mediated mast cell activation.
Nature communications (2016-10-28)
Kwok Ho Yip, Natasha Kolesnikoff, Nicholas Hauschild, Lisa Biggs, Angel F Lopez, Stephen J Galli, Sharad Kumar, Michele A Grimbaldeston
ZUSAMMENFASSUNG
Cross-linkage of the high-affinity immunoglobulin E (IgE) receptor (FcɛRI) on mast cells by antigen ligation has a critical role in the pathology of IgE-dependent allergic disorders, such as anaphylaxis and asthma. Restraint of intracellular signal transduction pathways that promote release of mast cell-derived pro-inflammatory mediators is necessary to dampen activation and restore homoeostasis. Here we show that the ligase Nedd4-2 and the adaptor Ndfip1 (Nedd4 family interacting protein 1) limit the intensity and duration of IgE-FcɛRI-induced positive signal transduction by ubiquitinating phosphorylated Syk, a tyrosine kinase that is indispensable for downstream FcɛRI signalosome activity. Importantly, loss of Nedd4-2 or Ndfip1 in mast cells results in exacerbated and prolonged IgE-mediated cutaneous anaphylaxis in vivo. Our findings reveal an important negative regulatory function for Nedd4-2 and Ndfip1 in IgE-dependent mast cell activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Phospho-LAT-(Tyr191-)Antikörper, Upstate®, from rabbit
Preise und Verfügbarkeit sind derzeit nicht verfügbar.