Reflectometric Determination of Nitrite in Meat Products After Reaction With Griess Reagent
Introduction
Nitrites, primarily in the form of sodium nitrite (E250) and potassium nitrite (E249), have been widely used as curing agents in meat production due to their multifunctional role in ensuring product quality and safety. They are directly responsible for the development of the characteristic flavor, stabilization of the cured red color, and inhibition of lipid oxidation. More importantly, nitrites exert antimicrobial effects, creating conditions unfavorable for the growth of spoilage organisms and foodborne pathogens, including Clostridium botulinum.1
Despite their functional importance, nitrites are associated with toxicological concerns. Under certain conditions, they can contribute to the formation of N-nitroso compounds, a group of substances classified by the International Agency for Research on Cancer (IARC) as probable human carcinogens.1 The regulation of nitrite use has therefore focused on achieving a balance between ensuring microbiological safety and minimizing the formation of harmful nitrosamines. This balance is reflected in strict maximum levels set by the European Union [Regulation (EC) No 1333/2008 and its subsequent amendments],2 and other international authorities which continue to guide the use of nitrites in cured and dried meat products.
Given these safety considerations, accurate monitoring of nitrite levels in meat products is essential. Reflectometric determination using the Griess method provides a simple, sensitive, and reliable analytical tool for this purpose, ensuring compliance with food safety standards while supporting routine quality control in the meat industry.
Experimental
This application note describes the rapid quantification of nitrite in meat products using the RQflex® 20 alongside the Reflectoquant® Nitrite Test (1.16973).
Method
In the presence of an acidic buffer nitrite ions react with an aromatic amine to form a diazonium salt, which in turn reacts with N-(1- naphthyl)-ethylene-diamine to form a red-violet azo dye that is determined reflectometrically.
Measuring Range
- Test kit: 0.5 - 25.0 mg/L NO2– (0.2 - 7.6 mg/L NO2–N)
- Method: 1.5 - 75 mg/kg NO2–
Applicable Sample
Chopped meat
Influence of Foreign Substances
Foreign substances in the sample solution can
- increase the measurement value because of an amplification of the reaction
- lower the measurement value because of a prevention of the reaction
A quantification of these effects is stated in tabular form in the respective package insert for the most important foreign ions and substances. The tolerance limits have been determined for the individual ions and substances; they may not be evaluated cumulatively.
In the case of samples with a complex, in many cases inexactly known composition (matrix) it is particularly difficult to estimate the potential influence of the foreign substances on the analysis (matrix effect). The following instructions describe a method by means of which the user can test whether a matrix effect is present or not.
Reagents, Instruments and Materials
Test/Reagents Kit
- Reflectoquant® Nitrite Test (1.16973)
Instrument(s) & Devices
- Reflectometer RQflex® 20, Reflectoquant® (1.17246)
Other Reagents and Accessories
- MQuant® Nitrite Test 2 - 80 mg/L NO2- (0.6 - 24 mg/L NO2-N) (1.10007)
- Distilled water (e.g., from a suitable Milli-Q® system) or water for analysis (1.16754)
- Balance
- Beaker
- Blender
- Heating plate
- Folded filter
Products for AQA
- RQcheck set for RQflex® 20 Reflectometer (1.17247)
- Recalibration Set for RQflex® 20 Reflectometer (1.16954)
- Nitrite standard solution Certipur®, 1000 mg/L NO2- (1.19899)
Analytical Procedure
Sample Preparation
- Weigh approximately 50 g chopped meat exactly into a beaker, add 150 ml distilled water and homogenize in a blender.
- Heat this mixture to 80 °C and filter through a folded filter. Allow the solution to cool down to room temperature before starting the analysis.
- Check the nitrite content with the MQuant® Nitrite Test. Samples containing more than 25.0 mg/L NO2- must be diluted with distilled water.
Measurement
- Press the START button of the reflectometer and - this is imperative - at the same time immerse both reaction zones of the test strip in the pretreated sample (15 - 30 °C) for 2 seconds.
- Carefully allow excess liquid to run off via the long edge of the strip onto an absorbent paper towel.
- Immediately insert the strip all the way into the strip adapter with the reaction zones facing the display.
- After the end of the reaction time (15 sec), read off the result from the display in mg/L NO2-. The result is automatically stored.
Notes on the measurement:
For detailed operating procedures, consult the operating instructions for RQflex® instrument and the package insert of the Reflectoquant® Nitrite Test.
Calculation
Nitrite content [mg/kg] = Measured value [mg/L] x 150 [mL] /Weight of sample [g]
Analytical quality assurance
Analytical quality assurance (AQA) is recommended before each measurement series. Check the instrument using the RQcheck. If RQcheck failed, perform a recalibration using the recalibration set and repeat the RQcheck. For more details see RQflex® 20 manual.
To check test strips, test reagent, measurement device, and handling (recommended before each measurement series): Dilute the nitrite standard solution with distilled water to 10.0 mg/L NO2- and analyze as described in the section measurement.
Results and Discussion
Here in this study, four different samples of chopped meat were used as test samples for measurement and comparison. The results were verified using ion chromatography as orthogonal method to the Reflectoquant® test kit.
Sample | Reflectoquant® [mg/kg] | IC [mg/kg] |
|---|---|---|
1 | 0.8 | 0.9 |
2 | 6.0 | 6.8 |
3 | 3.2 | 2.8 |
4 | 2.2 | 2.0 |
Conclusion
Rapid testing of nitrite content in meat products can be performed with Reflectoquant® Nitrite Test (1.16973) with RQflex® 20 reflectometer.
Orthogonal method using ion chromatography has provided evidence that experimental results from both methods have differences within ± 10 %.