Skip to Content
Merck

Skip To

BDPH9BCC2A67

Bide Pharmatech

4-Fluoro-3-(trifluoromethyl)benzoyl chloride

Synonym(s):

4-fluoro-3-(trifluoromethyl)benzoyl chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H3O1Cl1F4
CAS Number:
Molecular Weight:
226.56
MDL number:
Product BDPH9BCC2A67 is not currently sold in your country.


Inchi Code

1S/C8H3ClF4O/c9-7(14)4-1-2-6(10)5(3-4)8(11,12)13/h1-3H

Inchi Key

BUDISZQHCHGLJW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Colorless to Yellow Liquid



Still not finding the right product?


hstatements

signalword

Danger

pictograms

Corrosion


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library