Skip to Content
Merck

BDPH9BCD1C19

Bide Pharmatech

2,6,10-Trimethyl-2,6,10-triazaundecane

Synonym(s):

N~1~-[3-(dimethylamino)propyl]-N~1~,N~3~,N~3~-trimethyl-1,3-propanediamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H27N3
CAS Number:
Molecular Weight:
201.36
MDL number:
Product BDPH9BCD1C19 is not currently sold in your country.


Inchi Code

1S/C11H27N3/c1-12(2)8-6-10-14(5)11-7-9-13(3)4/h6-11H2,1-5H3

Inchi Key

SKCNNQDRNPQEFU-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Colorless to yellow liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library