Skip to Content
Merck

Skip To

52182

0.1 M Citrate pH 5.0 – 30% PEG 6000 solution

Synonym(s):

PEG Grid Screening Solution 32/Kit-No 36436

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352200


pH

5.0±0.2 ( in neat)

density

1.06 g/mL at 20 °C

storage temp.

2-8°C



Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
704040-5G04061832545905
704040-1G04061825972190

Questions

Reviews

No rating value

Active Filters