Adsorbenti al carbonio
Abbiamo più di 50 anni di esperienza nella ricerca e nello sviluppo di adsorbenti in carbonio e resina. La nostra linea completa di adsorbenti speciali al carbonio comprende setacci molecolari di carbonio (Carboxen® e Carbosieve™), carbonio polimerico sferico grafitizzato (Graphsphere™) e nero di carbonio grafitizzato (Carbotrap® e Carbopack™). Offriamo anche kit di campionatura, un modo economico per valutare diversi dei nostri adsorbenti speciali al carbonio.
Prodotti
Filtri popolari
Risorse correlate
- Application Note: Rhodium and Palladium Metal Scavenging with Carboxen® Synthetic Carbon Adsorbents
The ICH-Q3D-R2 guidelines on permissible elemental impurity levels in drug products require a metal scavenging clean-up step for many active pharmaceutical ingredients (APIs). Organometallic catalysts based on palladium and rhodium are commonly used as process catalysts for the synthesis of APIs. These metals fall under Class-2B of the ICH-Q3D-R2 guidelines, with an oral consumption limit of 10 ppm.
- Application Note: Surface Chemistry of Synthetic Carbon Adsorbents and their Application in the Adsorption of Amines
Activated carbons are widely used for purification and separation applications, such as purification of permanent gases, removal of volatile organic compounds from streams of gases and liquids, hydrocarbon vapor recovery, purification of active pharmaceutical ingredients, and preparing electrode materials in energy storage devices.
- Application Note: Carboxen® Synthetic Carbon Adsorbents for Host Cell Protein Removal During Monoclonal Antibody Purification
The demand for mAbs is steadily increasing while the depletion of host cell proteins (HCP) in the manufacturing of mAbs remains a challenge. HCPs can represent an immune response risk after the administration of mAbs if not reduced to appropriate levels.
- White Paper: Carboxen® Synthetic Carbon Adsorbents for Industrial Water Purification
This paper summarizes a selection of field studies that utilized Carboxen® 563 Synthetic Carbon for the purification of chemical wastewater and contaminated groundwater.
- Brochure: Specialty Carbon Adsorbents Advanced Materials for Industrial Purifications and Separations
Our specialty carbon adsorbents are highly engineered materials, manufactured from highly pure synthetic polymers. These products are then activated by physical means, avoiding the use of chemicals to activate. This differentiates them from common non-specific, low-tech carbon black and activated carbon adsorbents.
- Brochure: Resins Selection Guide
Low pressure liquid chromatography (LPLC) is the oldest of chromatographic techniques, originally performed with open columns that use gravity to move the sample through the packing bed. Now, pumps and other instrumentation have been incorporated into LPLC and superior packing media have been developed specifically for this versatile technology.
Carboxen® Adsorbents
Il Gli adsorbenti al carbonio sintetici Carboxen® sono resine derivatizzate altamente ingegnerizzate. Hanno una struttura di pori affusolata, da macro a meso a microporosa. I pori affusolati migliorano l'efficienza cinetica e termodinamica dell'adsorbente. Le particelle sono sferiche per un'efficienza ottimale del letto impaccato. Le resine derivatizzate altamente ingegnerizzate hanno un'eccellente resistenza all'attrito che consente di raggiungere portate elevate senza accumuli indesiderati di contropressione. Sono state sviluppate per resistere a pressioni fino a 16.000 psi. Sono stabili fino a 400 ˚C e hanno chimiche di superficie personalizzabili per l'analisi di target specifici. L'elevata purezza, la forma sferica e le eccezionali proprietà meccaniche li rendono adatti all'uso in cromatografia preparativa e di processo. I materiali sono conservati come perle secche e scorrono come un liquido, facilitando la manipolazione e il confezionamento delle colonne cromatografiche rispetto alle resine umide.
Il pH dei carboni sintetici Carboxen® può essere personalizzato da 2,5 a 10,5. Essendo prodotti da una fonte sintetica, i Carboxen® Synthetic Carbons sono intrinsecamente senza metalli pesanti e altri contenuti inorganici presenti negli adsorbenti di origine naturale. Gli adsorbenti al carbone sintetico Carboxen® sono scalabili, il che significa che è possibile produrre quantità di tonnellate di questi adsorbenti in maglia 20/40, 20/45 o 30/45. I prodotti Carboxen® di dimensioni più piccole sono offerti su scala ridotta.
Queste resine derivatizzate apportano valore a un'ampia gamma di applicazioni di purificazione per la rimozione di impurità di piccole e grandi molecole. Trovano impiego sia nella purificazione in fase gassosa che liquida. Alcuni esempi includono la rimozione di impurità ad alto rischio nelle purificazioni biochimiche come le proteine delle cellule ospiti, la rimozione di metalli pesanti tossici, la purificazione di metalli clorurati, la purificazione di API e la rimozione di sostanze estraibili e lisciviabili.
La NASA si è affidata a loro per diverse missioni, compreso l'attuale utilizzo sulla Stazione Spaziale Internazionale.
Visualizza l'articolo completo qui: https://www.sigmaaldrich.com/US/it/life-science/supelco/supelco-products-in-space
Le proprietà materiali degli otto Carboxen® Synthetic Carbon disponibili su scala bulk sono riportate nella tabella seguente.
Carboxen® Synthetic Carbon Material Properties
| Area superficiale (m2/g) | Ph della superficie (ASTM D6851) | 1=più idrofilo 10=più idrofobo | Struttura del poro (%) | Durezza Ball Pan (ASTM D3802) | |||
| Micro (< 20A) | Meso (20-500A) | Macro (> 500A) | |||||
| Carboxen® 563 | 500 | 6.8 | 7 | 38 | 24 | 38 | 99.8 |
| Carboxen® 564 | 500 | 8.7 | 8 | 47 | 26 | 27 | 99.8 |
| Carboxen® 569 | 500 | 8.6 | 9 | 45 | 32 | 23 | 99.4 |
| Carboxen® 572 | 850 | 9.5 | 4 | 48 | 23 | 29 | 99.1 |
| Carboxen® 1005 | 750 | 8.0 | 10 | 47 | 26 | 27 | 99.6 |
| Carboxen® 1032 | 800 | 3.0 | 1 | 49 | 0 | 51 | 98.6 |
| Carboxen® 1034 | 1200 | 10.5 | 2 | 52 | 3 | 45 | 98.9 |
Carbosieve® Adsorbents
Il Gli adsorbenti di carbosieve® sono setacci molecolari di carbonio con pori non conici. Con una struttura microporosa, offrono una forza adsorbente molto forte. Gli adsorbenti Carbosieve® offrono un'eccellente ritenzione di analiti piccoli e volatili che la maggior parte degli altri adsorbenti ha difficoltà a trattenere.
Nero di carbonio grafitizzato
Nero di carbonio grafitizzato a base di Carbopack™ e Gli adsorbenti Carbotrap® sono adsorbenti non porosi e non specifici che presentano un'elevata omogeneità superficiale. L'elevata purezza di questi adsorbenti garantisce un efficace desorbimento dell'analita di interesse. Presentano caratteristiche superficiali idrofobiche e, pertanto, possono essere efficacemente utilizzati per intrappolare i composti organici in flussi umidi. I composti intrappolati possono essere desorbiti mediante solvente o desorbimento termico con un'efficienza di desorbimento del 100%. Questi adsorbenti offrono un'eccellente stabilità termica, garantiscono un bleed minimo alle temperature di desorbimento termico e impediscono un'elevata caduta di pressione.
Graphsphere™ Adsorbenti
Il carbonio polimerico sferico grafitizzato (Graphsphere™) ha una struttura unica con un nucleo amorfo e un guscio esterno grafitico di spessore controllato. Il processo di grafitizzazione produce una superficie altamente pura con proprietà di adsorbimento e desorbimento uniche. Le interazioni superficiali dipendono esclusivamente dalle forze di dispersione (London). I materiali sono:
- Sferici (offrono una migliore prestazione del letto impaccato rispetto alle particelle granulari)
- Duri e non friabili
- Utilizzati per molecole con dimensioni dell'analita relative a C5-C12 n-alcani
- Idrofobico (può essere utilizzato in ambienti ad alta umidità)
Graphsphere™ adsorbents offrono una forza adsorbente relativa più debole rispetto agli adsorbenti a setaccio molecolare di carbonio e una forza adsorbente relativa simile rispetto agli adsorbenti a nero di carbonio grafitizzato.