サイズを選択してください
100 EA
¥9,660
¥9,660
単価¥13,800割引30%カートの在庫状況を確認する
この商品について
NACRES:
NB.01
UNSPSC Code:
41121522
スキップする
製品名
Reagent Reservoir, capacity 100 mL, PVC, non-sterile, pack of 100 (bulk packed)
material
PVC
V-bottom bottom
sterility
non-sterile
packaging
pack of 100 (bulk packed)
manufacturer/tradename
Heathrow Scientific 120650
capacity
100 mL
類似した製品をお探しですか? 訪問 製品比較ガイド
1 of 4
当該品目 | HS120646 | HS120645 | HS120638 |
|---|---|---|---|
| manufacturer/tradename Heathrow Scientific 120650 | manufacturer/tradename Heathrow Scientific 120646 | manufacturer/tradename Heathrow Scientific 120645 | manufacturer/tradename Heathrow Scientific 120638 |
| material PVC , V-bottom bottom | material PVC , V-bottom bottom | material V-bottom bottom, polystyrene | material V-bottom bottom, polystyrene |
| capacity 100 mL | capacity 50 mL | capacity 50 mL | capacity 10 mL |
| packaging pack of 100 (bulk packed) | packaging pack of 100 (packed as 5 ea) | packaging pack of 100 (bulk packed) | packaging pack of 100 (bulk packed) |
| sterility non-sterile | sterility sterile; electron beam irradiated | sterility non-sterile | sterility non-sterile |
Features and Benefits
- Trough design with a “v” shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
- Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
- Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
- Designed to stay flat on the benchtop and not warp
- Sealed in easy-to-open bags. No scissors needed to open
- Disposable
- Design Patent Pending
General description
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures. Graduation markingson the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents. Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping.