サイズを選択してください
| サイズ/SKU | 在庫状況 | 単価 |
|---|
この商品について
material
assorted colors , cardboard
feature
to hold 100 x cryovials
packaging
pkg of 5 ea
manufacturer/tradename
MTC™ Bio R2700-A
L × W × H
133 mm × 133 mm × 51 mm
General description
Features and Benefits
- Attached Hinged Lids
- Foolproof identification
- 5 1/4 x 5 1/4 x 2 inches
- Standard size, for existing racks
- Available in 6 colors
- Individual or rainbow packs
- For 81 or 100 tubes
- 9×9 or 10×10 format
- Alpha-numeric coding
- Top, sides, and bottom
Legal Information
1 of 1
当該品目 | |||
|---|---|---|---|
| description cardboard, assorted colors, to hold, 100 x cryovials, pkg of 5 ea | description cardboard, purple, to hold, 100 x cryovials, pkg of 5 ea | description cardboard, green, to hold, 100 x cryovials, pkg of 5 ea | description cardboard, orange, to hold, 100 x cryovials, pkg of 5 ea |
| manufacturer/tradename MTC™ Bio R2700-A | manufacturer/tradename MTC™ Bio R2700-P | manufacturer/tradename MTC™ Bio R2700-G | manufacturer/tradename MTC™ Bio R2700-O |
| feature to hold 100 x cryovials | feature to hold 100 x cryovials | feature to hold 100 x cryovials | feature to hold 100 x cryovials |
| material assorted colors , cardboard | material purple , cardboard | material cardboard , green | material orange , cardboard |
| packaging pkg of 5 ea | packaging pkg of 5 ea | packaging pkg of 5 ea | packaging pkg of 5 ea |
| L × W × H 133 mm × 133 mm × 51 mm | L × W × H 133 mm × 133 mm × 51 mm | L × W × H 133 mm × 133 mm × 51 mm | L × W × H 133 mm × 133 mm × 51 mm |
Still not finding the right product?
Explore all of our products under MTC™ Bio FlipTop™ Freezer Box With Hinged Lid