スキップする
由来生物
rabbit (Monoclonal)
品質水準
リコンビナント
expressed in HEK 293 cells
結合体
unconjugated
抗体製品の状態
affinity isolated antibody
抗体製品タイプ
primary antibodies
クローン
23GB5470, recombinant monoclonal
フォーム
buffered aqueous glycerol solution
分子量
73 kDa
交差性
human
包装
vial of 1.5 mL
強化検証
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
濃度
1 mg/ml
テクニック
flow cytometry: 1:2000
western blot: 1:1,000-1:5,000
アイソタイプ
IgG
Ensembl |ヒトアクセッション番号
NCBIアクセッション番号
UniProtアクセッション番号
環境により配慮した代替製品カテゴリ
輸送温度
wet ice
保管温度
−20°C
ターゲットの翻訳後修飾
unmodified
遺伝子情報
human ... FZD8(8325)
関連するカテゴリー
詳細
This intronless gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. This gene is highly expressed in two human cancer cell lines, indicating that it may play a role in several types of cancer. The crystal structure of the extracellular cysteine-rich domain of a similar mouse protein has been determined. [provided by RefSeq, Jul 2008]
We are committed to bringing you Greener Alternative Products. This product is a Design for Sustainability (DfS) developed product, which belongs to one of the four categories of greener alternatives. Click here to view its DfS scorecard.
免疫原
A synthesized peptide corresponding to a sequence within amino acids 640-694 of human Frizzled 8 [NP_114072.1].
物理的形状
Solution in PBS (pH 7.4) containing 50% glycerol, and 0.02% sodium azide.
その他情報
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user.
適切な製品が見つかりませんか。
製品選択ツール.をお試しください
関連コンテンツ
ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.
製品に関するお問い合わせはこちら(テクニカルサービス)
