由来生物
rabbit (Monoclonal)
品質水準
リコンビナント
expressed in HEK 293 cells
結合体
unconjugated
抗体製品の状態
affinity isolated antibody
抗体製品タイプ
primary antibodies
クローン
23GB5505, recombinant monoclonal
フォーム
buffered aqueous glycerol solution
分子量
52 kDa
交差性
rat, human
包装
vial of 1.5 mL
強化検証
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
濃度
1.5 mg/ml
テクニック
flow cytometry: 1:2000
western blot: 1:1,000-1:5,000
アイソタイプ
IgG
Ensembl |ヒトアクセッション番号
NCBIアクセッション番号
UniProtアクセッション番号
環境により配慮した代替製品カテゴリ
輸送温度
wet ice
保管温度
−20°C
ターゲットの翻訳後修飾
unmodified
遺伝子情報
human ... GABRA5(2558)
詳細
GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified. Transcript variants utilizing three different alternative non-coding first exons have been described. [provided by RefSeq, Jul 2008]
We are committed to bringing you Greener Alternative Products. This product is a Design for Sustainability (DfS) developed product, which belongs to one of the four categories of greener alternatives. Click here to view its DfS scorecard.
免疫原
A synthesized peptide corresponding to a sequence within amino acids 1-200 of human GABA A Receptor alpha 5 [NP_000801.1].
物理的形状
Solution in PBS (pH 7.4) containing 50% glycerol, and 0.02% sodium azide.
その他情報
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user.